Skip to Content
Merck
All Photos(4)

Key Documents

P5585

Sigma-Aldrich

Palmitic acid

BioXtra, ≥99%

Synonym(s):

1-Pentadecanecarboxylic acid, C16:0, Cetylic acid, Hexadecanoic acid, NSC 5030, PamOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOH
CAS Number:
Molecular Weight:
256.42
Beilstein:
607489
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

plant (vegetable)

grade

BioPerformance Certified

vapor pressure

10 mmHg ( 210 °C)

product line

BioXtra

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

ign. residue

≤0.1%

bp

271.5 °C/100 mmHg (lit.)

mp

61-62.5 °C (lit.)

solubility

chloroform: 0.5 M, clear, colorless

density

0.852 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCC(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)

InChI key

IPCSVZSSVZVIGE-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Palmitic acid has been used:
  • as a medium supplement in chorionic villous tissue culture form placenta[1]
  • as a component of hormone pellet for implantation in mice ovary[2]
  • for inducing expression of immunity-related GTPase M (IRGM) in human monocytic cell line, THP-1[3]

Biochem/physiol Actions

Palmitic acid (PA) is a component of membrane phospholipids (PL) and adipose triacylglycerols (TAG).[4] Elevated levels of palmitic acid contributes to the pathophysiology of atherosclerosis, type 2 diabetes mellitus, neurodegenerative diseases, obesity and cancer.[4][5] PA promotes apoptosis in the endothelial cell by modulating the p38 mitogen-activated protein kinase (MAPK) pathway and favors expression of TNF-α and reactive oxygen species accumulation.[6] PA promotes interleukin 8 (IL-8) synthesis in hepatocytes contributing to hepatic inflammation.[7] PA by interacting with toll-like receptor 4 (TLR4) induces inflammatory injury in cardiomyoctes.[8]
Palmitic acid is a straight-chain 16-carbon fatty acid. Diets rich in palmitic acid, along with lauric and myristic acids, are associated with increased serum levels of low-density lipoprotein cholesterol. Palmitic acid is the first fatty acid produced by lipogenesis. It is a negative regulator of acetyl-CoA carboxylase (ACC) and is implicated in fatty acid-induced insulin resistance.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biological and nutritional properties of palm oil and palmitic acid: effects on health
Mancini A, et al.
Molecules (Basel), 20(9), 17339-17361 (2015)
Lipidomic Analysis of alpha Synuclein Neurotoxicity Identifies Stearoyl CoA Desaturase as a Target for Parkinson Treatment
Fanning S, et al.
Molecular Cell (2018)
Palmitic acid induced vascular endothelial cell apoptosis contributes to the high severity of coronary artery disease in the Uygur population
Jiang H, et al.
Biomedical Research, 28(2), 17339-17361 (2017)
Palmitic acid: Physiological role, metabolism and nutritional implications
Carta G, et al.
Frontiers in Physiology, 8, 902-902 (2017)
Cangju Qinggan Jiangzhi Decoction Reduces the Development of NonAlcoholic Steatohepatitis and Activation of Kupffer Cells
Cheng Y, et al.
Cellular Physiology and Biochemistry, 48(3), 971-982 (2018)

Articles

Lipid Induced Insulin Resistance

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

Fatty acid synthesis supports cancer cell proliferation, essential for membrane generation, protein modification, and bioenergetics.

See All

Questions

1–6 of 6 Questions  
  1. I want to know the storage temperature after reconstituting the product in the solvent? Also clarify regarding the type of solvent to be used?

    1 answer
    1. Unfortunately, the stability of this product in solution is not determined.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. I am curious about the storage temperature of this product.

    1 answer
    1. As listed in the PROPERTIES section of the product page, this product may be stored at room temperature.

      Helpful?

  5. Can you tell me the storage temperature for this product?

    1 answer
    1. As stated in the PROPERTIES section of the product page, this product is stored at room temperature.

      Helpful?

  6. Is this product soluble only in chloroform? Did you know something about its solubility in ethanol or DMSO?

    1 answer
    1. Stock solutions of palmitic acid can be prepared in 200 proof ethanol or DMSO at 20 mg/mL. Stock solutions should be purged with inert gas, such as argon or nitrogen, prior to storage.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service