Merck
  • INPP4B overexpression is associated with poor clinical outcome and therapy resistance in acute myeloid leukemia.
INPP4B overexpression is associated with poor clinical outcome and therapy resistance in acute myeloid leukemia.
Leukemia (2015-03-05)
I Dzneladze, R He, J F Woolley, M H Son, M H Sharobim, S A Greenberg, M Gabra, C Langlois, A Rashid, A Hakem, N Ibrahimova, A Arruda, B Löwenberg, P J M Valk, M D Minden, L Salmena
ABSTRACT
In this study, we investigated the role of inositol polyphosphate-4-phosphatase, type-II (INPP4B) in acute myeloid leukemia (AML). We observed that AML patients with high levels of INPP4B (INPP4B(high)) had poor response to induction therapy, shorter event-free survival and shorter overall survival. Multivariate analyses demonstrated that INPP4B(high) was an independent predictor of poor prognosis, significantly improving current predictive models, where it outperformed conventional biomarkers including FLT3-ITD and NPM1. Furthermore, INPP4B(high) effectively segregated relative risk in AML patients with normal cytogenetics. The role of INPP4B on the biology of leukemic cells was assessed in vitro. Overexpression of INPP4B in AML cell lines enhanced colony formation potential, recapitulated the chemotherapy resistance observed in AML patients and promoted proliferation in a phosphatase-dependent, and Akt-independent manner. These findings reveal that INPP4B(high) has an unexpected role consistent with oncogenesis in AML, in contrast to its previously reported tumor-suppressive role in epithelial cancers. Overall, we propose that INPP4B is a novel prognostic biomarker in AML that has potential to be translated into clinical practice both as a disease marker and therapeutic target.
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 1 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture