Skip to Content
Merck
All Photos(1)

Key Documents

646563

Supelco

DL-Dithiothreitol solution

1 M in H2O

Synonym(s):

(±)-threo-1,4-Dimercapto-2,3-butanediol solution, DTT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O2S2
CAS Number:
Molecular Weight:
154.25
Beilstein:
1719757
MDL number:
UNSPSC Code:
12161504
PubChem Substance ID:
NACRES:
NA.25

form

liquid

reaction suitability

reagent type: reductant

concentration

1 M in H2O

density

1.04 g/mL at 20 °C
1.035 g/mL at 25 °C

SMILES string

O[C@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dithiothreitol (DTT) is used in proteomics applications to maintain sulfhydryl (-SH) groups in the reduced state and for quantitative reduction of disulfide (-S-S-) groups. It is a frequently used reagent in buffers due to its ability to reduce the oxidation state of a protein sample and preserves enzymatic activity. DTT is oxidized to the cyclic disulfide during the reduction of other disulfides in the solution.
Dithiothreitol is an effective reducing agent for protein analysis. The sealed-ampule format is convenient and easy to use when preparing samples for electrophoresis or other applications and avoids waste and loss of potency.

Application

DL-Dithiothreitol solution has been used:
  • in western blot analysis
  • as a supplement in lysis buffer to study its effects on sperm lysis
  • as a component of cold homogenization buffer, and chilled wash buffer for the isolation of nuclei from frozen rat tissues
  • in sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE), chromatography, modification of cysteine-containing compounds

Features and Benefits

Low toxicity and reduced odor compared to other thiol compounds.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Willem-Jan M Schellekens et al.
Critical care (London, England), 18(1), R28-R28 (2014-02-11)
Diaphragm weakness induced by prolonged mechanical ventilation may contribute to difficult weaning from the ventilator. Hypercapnia is an accepted side effect of low tidal volume mechanical ventilation, but the effects of hypercapnia on respiratory muscle function are largely unknown. The
Calista K L Ng et al.
Human molecular genetics, 24(11), 3163-3171 (2015-02-26)
mRNA decay is an essential and active process that allows cells to continuously adapt gene expression to internal and environmental cues. There are two mRNA degradation pathways: 3' to 5' and 5' to 3'. The DCPS protein is the scavenger
Kazuya Ishikawa et al.
Nature communications, 5, 5430-5430 (2014-11-13)
Pathogen effector proteins are delivered to host cells to suppress plant immunity. However, the mechanisms by which effector proteins function are largely unknown. Here we show that expression of XopP(Xoo), an effector of rice pathogen Xanthomonas oryzae pv. oryzae, in
Boseon Kim et al.
The EMBO journal, 34(13), 1801-1815 (2015-05-17)
Terminal uridylyl transferases (TUTs) function as integral regulators of microRNA (miRNA) biogenesis. Using biochemistry, single-molecule, and deep sequencing techniques, we here investigate the mechanism by which human TUT7 (also known as ZCCHC6) recognizes and uridylates precursor miRNAs (pre-miRNAs) in the
Maria de la Paz Celorio-Mancera et al.
Insect biochemistry and molecular biology, 61, 34-45 (2015-05-03)
More than half of the proteome from mandibular glands in caterpillars is represented by chemosensory proteins. Based on sequence similarity, these proteins are putative transporters of ligands to gustatory receptors in sensory organs of insects. We sought to determine whether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service