Merck
  • Lefetamine, a controlled drug and pharmaceutical lead of new designer drugs: synthesis, metabolism, and detectability in urine and human liver preparations using GC-MS, LC-MS(n), and LC-high resolution-MS/MS.
Lefetamine, a controlled drug and pharmaceutical lead of new designer drugs: synthesis, metabolism, and detectability in urine and human liver preparations using GC-MS, LC-MS(n), and LC-high resolution-MS/MS.
Analytical and bioanalytical chemistry (2015-01-13)
Carina S D Wink, Golo M J Meyer, Josef Zapp, Hans H Maurer
ABSTRACT
Lefetamine (N,N-dimethyl-1,2-diphenylethylamine, L-SPA) was marketed as an opioid analgesic in Japan and Italy. After being widely abused, it became a controlled substance. It seems to be a pharmaceutical lead for designer drugs because N-ethyl-1,2-diphenylethylamine (NEDPA) and N-iso-propyl-1,2-diphenylethylamine (NPDPA) were confiscated by the German police. In contrast to these derivatives, metabolism and detectability of lefetamine were not studied yet. Therefore, phase I and II metabolism should be elucidated and correlated to the derivatives. Also the detectability using the authors' standard urine screening approaches (SUSA) needed to be checked. As lefetamine was commercially unavailable, it had to be synthesized first. For metabolism studies, a high dose of lefetamine was administered to rats and the urine samples worked up in different ways. Separation and analysis were achieved by gas chromatography-mass spectrometry (GC-MS) and liquid chromatography-high resolution-tandem mass spectrometry (LC-HR-MS/MS). In accordance with NEPDA and NPDPA, the following metabolic steps could be proposed: N-oxidation, N-dealkylation, mono- and bis-hydroxylation of the benzene ring, and hydroxylation of the phenyl ring only after N-dealkylation. The di-hydroxy metabolites were conjugated by methylation of one hydroxy group, and hydroxy metabolites by glucuronidation or sulfation. All initial metabolites could also be detected in human liver preparations. After a therapeutic lefetamine dose, the bis-nor, bis-nor-hydroxy, nor-hydroxy, nor-di-hydroxy metabolites could be detected using the authors' GC-MS SUSA and the nor-hydroxy-glucuronide by the LC-MS(n) SUSA. Thus, an intake of lefetamine should be detectable in human urine assuming similar pharmacokinetics.
Sigma-Aldrich
Chloroform, suitable for HPLC
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Chloroform, JIS 300, ≥99.0%, for residue analysis
Sigma-Aldrich
Oxalic acid dihydrate, JIS special grade, 99.5-100.2%
Sigma-Aldrich
Oxalic acid, SAJ first grade, ≥97.0%
Sigma-Aldrich
Chloroform, SAJ super special grade, ≥99.0%
Sigma-Aldrich
Chloroform, SAJ first grade, ≥99.0%, contains 0.4-0.8% ethanol
Sigma-Aldrich
Chloroform, JIS special grade, ≥99.0%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Oxalic acid dihydrate, SAJ first grade, 99.0-101.0%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Supelco
Chloroform, analytical standard
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
Oxalic acid dihydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (manganometric)
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Oxalic acid dihydrate, ACS reagent, ≥99%