Merck
  • A degradation study of cefepime hydrochloride in solutions under various stress conditions by TLC-densitometry.
A degradation study of cefepime hydrochloride in solutions under various stress conditions by TLC-densitometry.
Biomedical chromatography : BMC (2014-07-22)
Monika Dąbrowska, Małgorzata Starek, Jan Krzek, Elżbieta Papp, Piotr Król
ABSTRACT
A rapid, accurate and sensitive thin-layer chromatography (TLC) method with densitometric detection has been developed and validated for the determination of cefepime in pharmaceuticals. Chromatographic separation was achieved on a silica gel TLC F254 plates with a mobile phase consisting of ethanol-2-propanol-glacial acetic acid 99.5%-water (4:4:1:3, v/v). Densitometric detection was carried out at wavelength of 266 nm in reflectance/absorbance mode. The validation of the method was found to be satisfactory with high accuracy (from 99.24 to 101.37%) and precision (RSD from 0.06 to 0.36%). Additionally, the stability of cefepime in solution was investigated, including the effect of pH, temperature and incubation time. Favorable retention parameters (Rf , Rs, α) were obtained under the developed conditions, which guaranteed good separation of the studied components. The degradation process of cefepime hydrochloride was described by kinetic and thermodynamic parameters (k, t0.1 , t0.5 and Ea ). Moreover, the chemical properties of degradation products were characterized by the Rf values, absorption spectra, HPLC-MS/MS and TLC-densitometry analysis. As the method could effectively separate the active substance from its main degradation product (1-methylpyrrolidine), it can be employed as a method to indicate the stability of this drug.
Product Description
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Anti-KDR/Flk-1/VEGFR2 Antibody, clone CH-11, clone CH-11, Upstate®, from mouse
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
1-Methylpyrrolidine, ≥98.0% (GC)
Sigma-Aldrich
N-Methylpyrrolidine, 97%