Merck
Inhibition of monoacylglycerol lipase reduces nicotine withdrawal.
British journal of pharmacology (2014-09-27)
P P Muldoon, J Chen, J L Harenza, R A Abdullah, L J Sim-Selley, B F Cravatt, M F Miles, X Chen, A H Lichtman, M I Damaj
ABSTRACT
Abrupt discontinuation of nicotine, the main psychoactive component in tobacco, induces a withdrawal syndrome in nicotine-dependent animals, consisting of somatic and affective signs, avoidance of which contributes to drug maintenance. While blockade of fatty acid amide hydrolase, the primary catabolic enzyme of the endocannabinoid arachidonoylethanolamine (anandamide), exacerbates withdrawal responses in nicotine-dependent mice, the role of monoacylglycerol lipase (MAGL), the main hydrolytic enzyme of a second endocannabinoid 2-arachidonylglycerol (2-AG), in nicotine withdrawal remains unexplored. To evaluate the role of MAGL enzyme inhibition in nicotine withdrawal, we initially performed a genetic correlation approach using the BXD recombinant inbred mouse panel. We then assessed nicotine withdrawal intensity in the mouse after treatment with the selective MAGL inhibitor, JZL184, and after genetic deletion of the enzyme. Lastly, we assessed the association between genotypes and smoking withdrawal phenotypes in two human data sets. BXD mice displayed significant positive correlations between basal MAGL mRNA expression and nicotine withdrawal responses, consistent with the idea that increased 2-AG brain levels may attenuate withdrawal responses. Strikingly, the MAGL inhibitor, JZL184, dose-dependently reduced somatic and aversive withdrawal signs, which was blocked by rimonabant, indicating a CB1 receptor-dependent mechanism. MAGL-knockout mice also showed attenuated nicotine withdrawal. Lastly, genetic analyses in humans revealed associations of the MAGL gene with smoking withdrawal in humans. Overall, our findings suggest that MAGL inhibition maybe a promising target for treatment of nicotine dependence.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, JIS first grade, 94.8-95.8%
Sigma-Aldrich
Ethanol, JIS special grade, 94.8-95.8%
Sigma-Aldrich
Mecamylamine hydrochloride
Nicotine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, for residue analysis
Supelco
Mycophenolic acid, analytical standard
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology