Skip to Content
Merck
All Photos(1)

Key Documents

M5255

Sigma-Aldrich

Mycophenolic acid

from Penicillium brevicompactum, ≥98% (HPLC), powder, IMP dehydrogenase inhibitor

Synonym(s):

6-(1,3-Dihydro-7-hydroxy-5-methoxy-4-methyl-1-oxoisobenzofuran-6-yl)-4-methyl-4-hexanoic acid, 6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl)-4-methyl-4-hexenoic acid, NSC 129185

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20O6
CAS Number:
Molecular Weight:
320.34
Beilstein:
1295848
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Product Name

Mycophenolic acid, ≥98%

biological source

Penicillium brevicompactum

Quality Level

Assay

≥98%

color

white to yellow-white

mp

<143.0 °C

solubility

methanol: 49.00-51.00 mg/mL, clear, colorless to faintly yellow

Mode of action

enzyme | inhibits

originator

Novartis

storage temp.

2-8°C

SMILES string

COc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)CCC(O)=O

InChI

1S/C17H20O6/c1-9(5-7-13(18)19)4-6-11-15(20)14-12(8-23-17(14)21)10(2)16(11)22-3/h4,20H,5-8H2,1-3H3,(H,18,19)/b9-4+

InChI key

HPNSFSBZBAHARI-RUDMXATFSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Mycophenolic acid (MPA), produced by Penicillum brevi-compactum, is a selective inhibitor of inosine monphosphate dehydrogenase, thus inhibiting DNA synthesis in T and B lymphocytes. It has also been shown to act as an immunosuppressive agent, and as an inducer of monocyte differentiation and apoptosis in human lymphoid and monocytic cell lines. As a selection agent, MPA is used for transfected animal cells expressing the E. Coli gene for xanthine-guanine phosphoribosyl transferase, and is recommended for use at 25μg/mL.

Biochem/physiol Actions

Mode of Action: This product acts by suppressing the cytokine-induced nitric oxide production, inhibiting early stage biosynthesis of purine nucleotides and as a specific inhibitor of IMP dehydrogenase.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Caution

As supplied, this product should be stored desiccated at 2-8°C, and is stable for 5 years when stored properly.

Preparation Note

Mycophenolic acid is soluble in methanol at 50 mg/mL, yielding a colorless to faint yellow solution, as well as chloroform, dichloromethane, ethanol and .1 N NaOH. After reconstitution, the recommendation is to sterilize via filtration thorugh a 0.22 μm pore-size filter, aliquot, and freeze at -20°C.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2 - Repr. 1B - STOT RE 1 Oral

Target Organs

Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guste Urbonaite et al.
Communications biology, 4(1), 822-822 (2021-07-02)
Stochastic gene expression leads to inherent variability in expression outcomes even in isogenic single-celled organisms grown in the same environment. The Drop-Seq technology facilitates transcriptomic studies of individual mammalian cells, and it has had transformative effects on the characterization of
Natacha S Ogando et al.
Transplant international : official journal of the European Society for Organ Transplantation, 35, 10369-10369 (2022-07-12)
Kidney transplant recipients (KTRs) are at increased risk for a more severe course of COVID-19, due to their pre-existing comorbidity and immunosuppression. Consensus protocols recommend lowering immunosuppression in KTRs with severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection, but the
Christine E Staatz et al.
Clinical pharmacokinetics, 50(12), 759-772 (2011-11-18)
This review seeks to summarize the available data about Bayesian estimation of area under the plasma concentration-time curve (AUC) and dosage prediction for mycophenolic acid (MPA) and evaluate whether sufficient evidence is available for routine use of Bayesian dosage prediction
Ping Liu et al.
Current genetics, 66(4), 813-822 (2020-04-02)
Slowing down aging-associated accumulation of molecular damage or its prevention represents a promising therapeutic paradigm to combat aging-related disease and death. While several chemical compounds extend lifespan in model organisms, their mechanism of action is often unknown, reducing their therapeutic
Amy Tarangelo et al.
Life science alliance, 5(4) (2022-01-26)
Nucleotide synthesis is a metabolically demanding process essential for DNA replication and other processes in the cell. Several anticancer drugs that inhibit nucleotide metabolism induce apoptosis. How inhibition of nucleotide metabolism impacts non-apoptotic cell death is less clear. Here, we

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service