Merck
Self-alignment of silver nanoparticles in highly ordered 2D arrays.
Nanoscale research letters (2015-04-18)
Ericka Rodríguez-León, Ramón Íñiguez-Palomares, Efraín Urrutia-Bañuelos, Ronaldo Herrera-Urbina, Judith Tánori, Amir Maldonado
ABSTRACT
We have synthesized silver nanoparticles in the non-polar phase of non-aqueous microemulsions. The nanocrystals have been grown by reducing silver ions in the microemulsion cylindrical micelles formed by the reducing agent (ethylene glycol). By a careful deposit of the microemulsion phase on a substrate, the micelles align in a hexagonal geometry, thus forming a 2D array of parallel strings of individual silver nanoparticles on the substrate. The microemulsions are the ternary system of anionic surfactant, non-polar solvent (isooctane), and solvent polar (ethylene glycol); the size of synthesized nanoparticles is about 7 nm and they are monodisperse. The study of the microstructure was realized by transmission electron microscopy, high-resolution technique transmission electron microscopy (HR-TEM), and Fourier processing using the software Digital Micrograph for the determination of the crystalline structure of the HR-TEM images of the nanocrystals; chemical composition was determined using the energy-dispersive X-ray spectroscopy. Addition technique polarizing light microscopy allowed the observation of the hexagonal phase of the system. This method of synthesis and self-alignment could be useful for the preparation of patterned materials at the nanometer scale.
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Silver nitrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Silver nitrate, puriss. p.a., ≥99.5% (AT)
Sigma-Aldrich
Silver nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
USP
Ethylene glycol, United States Pharmacopeia (USP) Reference Standard
Supelco
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, ReagentPlus®, ≥99.0% (titration)
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Silver nitrate, meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Silver nitrate, anhydrous, ≥99.999% trace metals basis
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Silver nitrate, tested according to Ph. Eur.
Sigma-Aldrich
Silver nitrate solution, 0.0282 M
Sigma-Aldrich
2,2,4-Trimethylpentane, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
2,2,4-Trimethylpentane, SAJ first grade, ≥98.0%
Sigma-Aldrich
Silver nitrate solution, 0.05 M
Sigma-Aldrich
2,2,4-Trimethylpentane, JIS special grade
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Docusate sodium salt, BioUltra, ≥99.0% (TLC)
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Docusate sodium salt, purum, ≥96.0% (TLC)