Skip to Content
Merck
All Photos(3)

Key Documents

S8157

Sigma-Aldrich

Silver nitrate

BioXtra, >99% (titration)

Synonym(s):

Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.31
Assay:
>99% (titration)
form:
solid
solubility:
H2O: 1 M at 20 °C, clear, colorless
Pricing and availability is not currently available.

vapor density

5.8 (vs air)

Quality Level

product line

BioXtra

Assay

>99% (titration)

form

solid

impurities

<0.005% Phosphorus (P)
<0.1% Insoluble matter

pH

5.0-7.0 (20 °C, 1 M in H2O)

mp

212 °C (dec.) (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): <0.05%
sulfate (SO42-): <0.05%

cation traces

Al: <0.0005%
Ca: <0.0005%
Cu: <0.0005%
Fe: <0.0005%
K: <0.005%
Mg: <0.0005%
NH4+: <0.05%
Na: <0.01%
Pb: <0.001%
Zn: <0.0005%

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver nitrate stains substance with brown-black color.[1] It is unstable at a temperature >100°C and loses its chromatographic properties.[2]

Application

Silver nitrate has been used:
  • in the synthesis of colloidal silver nanoparticles[3]
  • in von Kossa staining of differentiated osteogenically driven human mesenchymal stem cells,[4] osteoblasts and mineralized nodules[5]
  • in the approach of developing a new myelin silver stain[6]

Biochem/physiol Actions

Silver nitrate solution is originally used as an antiseptic agent and to treat burn wounds. Treatment with hypotonic silver nitrate solution might result in hyponatremia, an electrolyte abnormality.[1]

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A modified technique for high-resolution staining of myelin
Pistorio AL, et al.
Journal of Neuroscience Methods, 153(1), 135-146 (2006)
Trauma: Critical Care, 945-945 (2007)
Near-infrared optical imaging for monitoring the regeneration of osteogenic tissue-engineered constructs
Cowles EA, et al.
BioResearch Open Access, 2(3), 186-191 (2013)
Chromatography for the Analysis of Lipids, 28-28 (1993)
Cyclic AMP signaling in bone marrow stromal cells has reciprocal effects on the ability of mesenchymal stem cells to differentiate into mature osteoblasts versus mature adipocytes
Kao R, et al.
Endocrine, 42(3), 622-636 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service