Merck
  • Polyethyleneimine coating of magnetic particles increased the stability of an immobilized diglycosidase.
Polyethyleneimine coating of magnetic particles increased the stability of an immobilized diglycosidase.
Biotechnology and applied biochemistry (2014-04-05)
Marisol Minig, Laura S Mazzaferro, Agostina Capecce, Javier D Breccia
ABSTRACT
The diglycosidase, α-rhamnosyl-β-glucosidase, from Acremonium sp. DSM24697 was immobilized by adsorption and cross-linking onto polyaniline-iron (PI) particles. The immobilization yield and the immobilization efficiency were relatively high, 31.2% and 8.9%, respectively. However, the heterogeneous preparation showed lower stability in comparison with the soluble form of the enzyme in operational conditions at 60 °C. One parameter involved in the reduced stability of the heterogeneous preparation was the protein metal-catalyzed oxidation achieved by iron traces supplied from the support. To overcome the harmful effect, iron particles were coated with polyethyleneimine (PEI; 0.84 mg/g) previously for the immobilization of the catalyst. The increased stability of the catalyst was correlated with the amount of iron released from the support. Under operational conditions, the uncoated particles lost between 76% and 52% activity after two cycles of reuse, whereas the PEI-coated preparation reduced 45-28% activity after five cycles of reuse in the range of pH 5.0-10, respectively. Hence, polymer coating of magnetic materials used as enzyme supports might be an interesting approach to improve the performance of biotransformation processes.
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Supelco
Aniline, analytical standard
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sigma-Aldrich
Ammonium persulfate, JIS special grade, ≥98.0%
Sigma-Aldrich
Rutinose, ≥98.0% (HPLC)
Supelco
Hesperidin, analytical standard
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Hesperidin, ≥80% (HPLC)
Hesperidin, primary reference standard
Sigma-Aldrich
Ammonium persulfate, SAJ first grade, ≥95.0%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%
Hesperidin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium persulfate, free-flowing, Redi-Dri, reagent grade, 98%
Sigma-Aldrich
Ammonium persulfate, ACS reagent, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, reagent grade, 98%