Skip to Content
Merck
All Photos(4)

Key Documents

215589

Sigma-Aldrich

Ammonium persulfate

reagent grade, 98%

Synonym(s):

AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2S2O8
CAS Number:
Molecular Weight:
228.20
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38030421
PubChem Substance ID:
NACRES:
NA.55
Assay:
98%
grade:
reagent grade
form:
crystalline powder

grade

reagent grade

Quality Level

vapor density

7.9 (vs air)

Assay

98%

form

crystalline powder

reaction suitability

reagent type: oxidant

pH

1-2 (25 °C, 228 g/L)

SMILES string

N.N.OS(=O)(=O)OOS(O)(=O)=O

InChI

1S/2H3N.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h2*1H3;(H,1,2,3)(H,4,5,6)

InChI key

ROOXNKNUYICQNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for acrylamide gel polymerization.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

In vitro Transcription (IVT) and tRNA Binding Assay.
Schoenfelder SMK.
Bio-protocol, 4(18) (2014)
Solid state deoximation with ammonium persulfate-silica gel: Regeneration of carbonyl compounds using microwaves.
Varma RS and Meshram HM.
Tetrahedron Letters, 38(31), 5427-5428 (1997)
Electrical conductivity response of polypyrrole to acetone vapor: effect of dopant anions and interaction mechanisms.
Ruangchuay L, et al.
Synthetic Metals, 140(1), 15-21 (2004)
S Pino et al.
Clinical chemistry, 42(2), 239-243 (1996-02-01)
The chloric acid method is most commonly used to obtain accurate and reproducible measurements of iodine and remove interfering substances. Unfortunately chloric acid is a potential hazard, requiring an explosion-proof hood, among other precautions. We have developed a simple, convenient
J M Brewer
Science (New York, N.Y.), 156(3772), 256-257 (1967-04-14)
Ammonium persulfate, a common polymerizing agent for acrylamide gels, can inactivate yeast enolase and produce increased electrophoretic heterogeneity during disc electrophoresis in gels containing 8M urea. The use of riboflavin and light for polymerization or thioglycolate for removal of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service