Skip to Content
Merck
All Photos(1)

Documents

01-5610

Sigma-Aldrich

Aniline

SAJ first grade, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NH2
CAS Number:
Molecular Weight:
93.13
Beilstein:
605631
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.22 (185 °C, vs air)

vapor pressure

0.7 mmHg ( 25 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

1139 °F

expl. lim.

11 %

availability

available only in Japan

refractive index

n20/D 1.586 (lit.)

pH

8.8 (20 °C, 36 g/L)

bp

184 °C (lit.)

mp

−6 °C (lit.)

density

1.022 g/mL at 25 °C (lit.)

SMILES string

Nc1ccccc1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2

InChI key

PAYRUJLWNCNPSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

The acute toxicity of aniline involves its activation in vivo to 4-hydroxyaniline and the formation of adducts with hemoglobin. In erythrocytes, this is associated with the release of iron and the accumulation of methemoglobin and the development of hemolytic anemia and inflammation of the spleen. Tumor formation is often observed in the spleen on prolonged administration.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongli Ji et al.
Molecular plant-microbe interactions : MPMI, 28(5), 519-533 (2015-01-22)
The nonprotein amino acid β-aminobutyric acid (BABA) is known to protect plants against various pathogens. The mode of action is relatively diverse and specific in different plant-pathogen systems. To extend the analysis of the mode of action of BABA to
M Peluso et al.
Carcinogenesis, 21(2), 183-187 (2000-02-05)
The 'Mediterranean diet', a diet rich in cereals, fruit and vegetables, has been associated with lowering the risk of a variety of cancers of the digestive tract and the bladder. In a previous study, we showed that the high phenolic
Conghui Tang et al.
Journal of the American Chemical Society, 134(46), 18924-18927 (2012-11-08)
A novel and efficient copper-catalyzed azidation reaction of anilines via C-H activation has been developed. This method, in which the primary amine acts as a directing group by coordinating to the metal center, provides ortho azidation products regioselectively under mild
S A Vaziri et al.
Pharmacogenetics, 11(1), 7-20 (2001-02-24)
Arylamines such as 2-naphthylamine and 4-aminobiphenyl are suspected human bladder procarcinogens that require bioactivation to DNA-reactive species to exert their carcinogenic potential. The goals of the present study were (i) to assay for the presence of the arylamine acetyltransferases NAT1
J S Bus et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 25(8), 619-626 (1987-08-01)
Aniline and several structurally-related aromatic amines produce spleen tumours in rats given high doses of compound in 2-year bioassay studies. Evaluation of the pathogenesis of the splenic lesions and characterization of the disposition of radiolabelled aniline in animals suggests that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service