Merck
  • Analysis of 21 progestagens in various matrices by ultra-high-performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) with diverse sample pretreatment.
Analysis of 21 progestagens in various matrices by ultra-high-performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) with diverse sample pretreatment.
Analytical and bioanalytical chemistry (2014-09-28)
Shuang-Shuang Liu, Guang-Guo Ying, Shan Liu, Hua-Jie Lai, Zhi-Feng Chen, Chang-Gui Pan, Jian-Liang Zhao, Jun Chen
ABSTRACT
In this study, a highly sensitive and robust method using an ultra-high-performance liquid chromatography-tandem mass spectrometry combined with solid-phase extraction and ultrasonic extraction for pretreatment and silica gel purification steps has been developed for determination of 21 natural and synthetic progestagens in river surface water and sediments, and influents, effluents, and sludge from municipal wastewater treatment plants, and flush water and feces from swine farms. For the various matrices considered, the optimized method showed satisfactory performance with recoveries of 70-129 % (except AD, 5α-DHP, DPT, HPC), the limits of quantification below 2.30 ng/L for liquid samples and 2.59 ng/g for solid samples (except AD), and good linearity and reproducibility. This developed method was successfully applied in the analysis of progestagens in environmental samples from Liuxi Reservoir, Xintang municipal wastewater treatment plant, and Shunfeng swine farm in South China. Six analytes were detected at trace levels in surface water, effluent, and sediment samples. Seven analytes (0.7 (HPA)-35.1 ng/L (DGT)) were found in the influent samples and three analytes (5.6 (DGT)-11.8 ng/g (5α-DHP)) in the dewatered sludge samples. Moreover, 13 analytes were detected in swine farm, with high concentrations ranging from 23.8 ng/L (ET) to 5,024 ng/L (P) in flush water, and from 20.0 ng/g (MPA) to 1952 ng/g (P) in feces.
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Ammonium acetate solution, 50 % (w/v)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Divinylbenzene, technical grade, 55%
Supelco
Methanol, analytical standard