Skip to Content
Merck
All Photos(1)

Key Documents

01-4401

Sigma-Aldrich

Ammonium acetate solution

50 % (w/v)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONH4
CAS Number:
Molecular Weight:
77.08
Beilstein:
4186741
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

availability

available only in Japan

concentration

50 % (w/v)

SMILES string

CC([O-])=O.[H][N+]([H])([H])[H]

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Baron-Cohen et al.
Molecular psychiatry, 20(3), 369-376 (2014-06-04)
Autism affects males more than females, giving rise to the idea that the influence of steroid hormones on early fetal brain development may be one important early biological risk factor. Utilizing the Danish Historic Birth Cohort and Danish Psychiatric Central
Nazish Ahmed et al.
Tissue engineering. Part A, 20(15-16), 2224-2233 (2014-03-13)
One of the factors preventing clinical application of regenerative medicine to degenerative cartilage diseases is a suitable source of cells. Chondrocytes, the only cell type of cartilage, grown in vitro under culture conditions to expand cell numbers lose their phenotype
Lucie Nováková et al.
Analytica chimica acta, 853, 637-646 (2014-12-04)
The conditions for the analysis of selected doping substances by UHPSFC-MS/MS were optimized to ensure suitable peak shapes and maximized MS responses. A representative mixture of 31 acidic and basic doping agents was analyzed, in both ESI+ and ESI- modes.
Suzanne L Parker et al.
Journal of pharmaceutical and biomedical analysis, 105, 39-45 (2014-12-22)
A high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) method, using hydrophilic interaction liquid chromatography (HILIC) chromatography for the analysis of fosfomycin in human plasma and urine, has been developed and validated. The plasma method uses a simple protein precipitation using
Jane Kilcoyne et al.
Journal of agricultural and food chemistry, 63(20), 5083-5091 (2015-04-25)
Azaspiracids (AZAs) are marine biotoxins produced by dinoflagellates that can accumulate in shellfish, which if consumed can lead to poisoning events. AZA7-10, 7-10, were isolated from shellfish and their structures, previously proposed on the basis of only LC-MS/MS data, were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service