Merck
  • In situ synthesis of uniform Fe2O3/BiOCl p/n heterojunctions and improved photodegradation properties for mixture dyes.
In situ synthesis of uniform Fe2O3/BiOCl p/n heterojunctions and improved photodegradation properties for mixture dyes.
Dalton transactions (Cambridge, England : 2003) (2014-08-12)
Na Li, Xia Hua, Kai Wang, Yujian Jin, Jingjing Xu, Mindong Chen, Fei Teng
ABSTRACT
The xFe2O3/yBiOCl composites (xFe/yBi, x/y = 0/100, 5/100, 10/100, 20/100, 30/100 and 40/100 molar ratios) are prepared for the first time through an in situ hydrolysis method under hydrothermal conditions. The samples are characterized by X-ray diffraction (XRD), scanning electron microscopy (SEM), high-resolution transmission electron microscopy (HRTEM), selected area electron diffraction (SAED) and UV-visible diffuse reflectance spectroscopy (UV-DRS). The photodegradation performances of the xFe/yBi samples are investigated using a simulated industrial wastewater mixture containing both rhodamine B (RhB) and methyl orange (MO). The uniform Fe2O3 nanocubes are found to be well distributed on the BiOCl nanosheets. Moreover, the xFe/yBi photocatalysts exhibit unexpectedly higher efficiencies than bare BiOCl or Fe2O3 under ultraviolet light irradiation (λ ≤ 420 nm). Specifically, the degradation rates of the xFe/yBi samples at x/y = 5/100 and 10/100 are four times higher than that of bare BiOCl for MO-RhB mixed dye solutions. Their high photocatalytic activities are mainly attributed to the formation of stable p/n heterojunctions between Fe2O3 and BiOCl, which greatly improve the separation of photogenerated carriers. Importantly, the highly efficient and inexpensive xFe/yBi p/n heterojunctions are expected to be useful in practical industrial wastewater applications involving complicated toxic components.
Product Description
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bismuth(III) nitrate pentahydrate, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 1 M
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Bismuth(III) nitrate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Bismuth(III) nitrate pentahydrate, reagent grade, 98%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Bismuth(III) nitrate pentahydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%