Merck
  • Expression and function of fatty acid-binding protein 4 in epithelial cell of uterine endometrium.
Expression and function of fatty acid-binding protein 4 in epithelial cell of uterine endometrium.
Cell biology international (2015-01-13)
Qiuyuan Zhu, Yan Jin, Peng Wang, Hanzhi Wang, Bingjian Lu, Zhengping Wang, Minyue Dong
ABSTRACT
The aims of this study were to delineate the expression of fatty-acid binding protein (FABP) 4 in human uterine endometrium and its function in the regulation of proliferation, migration and invasion of epithelial cells. Immunohistochenistry, immunofluorence and Western blotting were used to determine the expression and cellular localization of FABP4 in endometrium and endometrial epithelial cell lines. Interference of small ribonuclear acid (siRNA) and specific FABP4 inhibitor were used to inhibit FABP4. The proliferation, migration and invasion of epithelial cells were evaluated with CCK-8 assay, wound-healing test and transwell analysis respectively. We found that FABP4 was expressed by epithelial cells of proliferative endometrium and epithelial and stromal cells of secrectory endometrium. Epithelial cell lines Ishikawa and RL-952 expressed FABP4 and this expression was decreased by FABP4 siRNA. FABP4 siRNA and specific FABP4 inhibition significantly decreased the proliferation, migration and invasion of epithelial cell lines. We concluded that FABP4 is functionally expressed in endometrial epithelium and is necessary for maintaining the cell function of epithelial cells of endometrium.
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
MISSION® esiRNA, targeting human FABP4
Sigma-Aldrich
FABP4 Inhibitor, The FABP4 Inhibitor, also referenced under CAS 300657-03-8, controls the biological activity of FABP4.
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O