Merck
  • HDL cholesterol performance using an ultracentrifugation reference measurement procedure and the designated comparison method.
HDL cholesterol performance using an ultracentrifugation reference measurement procedure and the designated comparison method.
Clinica chimica acta; international journal of clinical chemistry (2014-12-03)
Masakazu Nakamura, Shinji Yokoyama, Yuzo Kayamori, Hiroyasu Iso, Akihiko Kitamura, Tomonori Okamura, Masahiko Kiyama, Hiroyuki Noda, Kunihiro Nishimura, Michikazu Nakai, Isao Koyama, Mahnaz Dasti, Hubert W Vesper, Tamio Teramoto, Yoshihiro Miyamoto
ABSTRACT
Accurate high-density lipoprotein cholesterol (HDL-C) measurements are important for management of cardiovascular diseases. The US Centers for Disease Control and Prevention (CDC) and Cholesterol Reference Method Laboratory Network (CRMLN) perform ultracentrifugation (UC) reference measurement procedure (RMP) to value assign HDL-C. Japanese CRMLN laboratory (Osaka) concurrently runs UC procedure and the designated comparison method (DCM). Osaka performance of UC and DCM was examined and compared with CDC RMP. CDC RMP involved UC, heparin-MnCl₂ precipitation, and cholesterol analysis. CRMLN DCM for samples containing <200 mg/dl triglycerides involved 50-kDa dextran sulfate-MgCl2 precipitation and cholesterol determination. HDL-C regression equations obtained with CDC (x) and Osaka (y) were y=0.992x+0.542 (R(2)=0.996) for Osaka UC and y=1.004x-0.181 (R(2)=0.998) for DCM. Pass rates within ±1 mg/dl of the CDC target value were 91.9 and 92.1% for Osaka UC and DCM, respectively. Biases at 40 mg/dl HDL-C were +0.22 and -0.02 mg/dl for Osaka UC and DCM, respectively. Osaka UC and DCM were highly accurate, precise, and stable for many years, assisting manufacturers to calibrate products for clinical laboratories to accurately measure HDL-C for patients, calculate non-HDL-C, and estimate low-density lipoprotein cholesterol with the Friedewald equation.
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Manganese, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Manganese, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Manganese, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.0125mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
Manganese, rod, 10mm, diameter 4.0mm, cast, 99.5%
Manganese, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.009mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.003mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.003mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 98.7%
Manganese, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Manganese, rod, 50mm, diameter 6.35mm, cast, 99.5%
Manganese, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%
Manganese, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Manganese, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Manganese, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%