Skip to Content
Merck
All Photos(2)

Key Documents

GF99005193

Manganese

foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 98.7%

Synonym(s):

Manganese, MN000170

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mn
CAS Number:
Molecular Weight:
54.94
MDL number:
UNSPSC Code:
12141724
PubChem Substance ID:
NACRES:
NA.23

Assay

≥98.7%

form

foil

manufacturer/tradename

Goodfellow 990-051-93

resistivity

185 μΩ-cm, 20°C

bp

1962 °C (lit.)

mp

1244 °C (lit.)

density

7.3 g/mL at 25 °C (lit.)

SMILES string

[Mn]

InChI

1S/Mn

InChI key

PWHULOQIROXLJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirna Lechpammer et al.
Annals of neurology, 75(4), 608-612 (2014-03-07)
We followed a patient with manganese transporter deficiency due to homozygous SLC30A10 mutations from age 14 years until his death at age 38 years and present the first postmortem findings of this disorder. The basal ganglia showed neuronal loss, rhodanine-positive
Ebany J Martinez-Finley et al.
Free radical biology & medicine, 62, 65-75 (2013-02-12)
Manganese (Mn) is an essential dietary nutrient, but an excess or accumulation can be toxic. Disease states, such as manganism, are associated with overexposure or accumulation of Mn and are due to the production of reactive oxygen species, free radicals
Srinivasan V Narayanan et al.
Stroke, 46(6), 1626-1632 (2015-04-25)
Nuclear erythroid 2 related factor 2 (Nrf2) is an astrocyte-enriched transcription factor that has previously been shown to upregulate cellular antioxidant systems in response to ischemia. Although resveratrol preconditioning (RPC) has emerged as a potential neuroprotective therapy, the involvement of
Masakazu Nakamura et al.
Clinica chimica acta; international journal of clinical chemistry, 439, 185-190 (2014-12-03)
Accurate high-density lipoprotein cholesterol (HDL-C) measurements are important for management of cardiovascular diseases. The US Centers for Disease Control and Prevention (CDC) and Cholesterol Reference Method Laboratory Network (CRMLN) perform ultracentrifugation (UC) reference measurement procedure (RMP) to value assign HDL-C.
Marcus K Giacci et al.
PloS one, 9(8), e104565-e104565 (2014-08-12)
Red/near-infrared irradiation therapy (R/NIR-IT) delivered by laser or light-emitting diode (LED) has improved functional outcomes in a range of CNS injuries. However, translation of R/NIR-IT to the clinic for treatment of neurotrauma has been hampered by lack of comparative information

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service