Merck
  • Hydrolytic pathway of 5-fluorouracil in aqueous solutions for clinical use.
Hydrolytic pathway of 5-fluorouracil in aqueous solutions for clinical use.
Journal of pharmaceutical and biomedical analysis (2014-07-20)
Rémi Legay, Stéphane Massou, Joëlle Azéma, Robert Martino, Myriam Malet-Martino
ABSTRACT
The purpose of the study was to investigate the degradation pathway of 5-fluorouracil (FU) in the situation of commercial formulations for clinical use, namely FU dissolved in sodium hydroxide (NaOH) solutions or Tris buffer at pH 8.5-9. Combination of data from (19)F, (1)H and (13)C NMR and in some cases MS led to the identification of 8 and 13 FU degradation products in NaOH and Tris solutions respectively. In FU NaOH solutions, the first stage of FU degradation is a stereoselective hydration of the C5-C6 double bond leading to 5,6-dihydro-5-fluoro-6-hydroxyuracil, the cis stereoisomer being predominant relative to the trans. The second stage involves either a defluorination step with formation of fluoride ion and 5-hydroxyuracil or the cleavage of the N3-C4 bond giving the two diastereoisomeric 2-fluoro-3-hydroxy-3-ureidopropanoic acids. The subsequent N1-C6 bond breakdown of these compounds releases urea and 2-fluoro-3-oxopropanoic acid (FOPA) which in turn losses easily carbon dioxide leading to the formation of fluoroacetaldehyde (Facet). The degradation pathway in FU-Tris solutions is identical, except that Tris reacts with the aldehydes FOPA and Facet to form oxazolidine adducts stable at pH 8.5 but in equilibrium with the aldehyde forms at physiological pH, whereas the high reactivity of free aldehydes leads to numerous unidentified degradation compounds all in very low amounts. The FOPA diastereoisomeric adducts react with Facet to form four diastereoisomeric fused bicyclic five-membered ring compounds. Facet and FOPA are highly cardiotoxic. In Tris formulations, they are trapped as stable oxazolidine adducts which release the free aldehydes at physiological pH thus explaining the higher cardiotoxicity of FU in Tris solutions compared to that of FU in NaOH solutions.
Sigma-Aldrich
Sodium propionate, meets analytical specification of NF, 99-100.5% (ex dried subst.)
Sigma-Aldrich
Pyridinium chlorochromate, 98%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Malonic acid, ReagentPlus®, 99%
Sigma-Aldrich
Sodium propionate, FCC
Sigma-Aldrich
Sodium propionate, ≥99.0%, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sodium propionate, ≥99.0%
Supelco
Malonic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sodium propionate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Glycerol, JIS special grade, ≥99.0%
Supelco
Barbituric acid, for spectrophotometric det. of cyanide, ≥99.5%
Sigma-Aldrich
Chloroacetaldehyde solution, ~55 wt. % in H2O
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Chromium(III) acetylacetonate, 97%
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Chloroacetaldehyde solution, produced by Wacker Chemie AG, Burghausen, Germany, ≥45.0% in H2O (density determination)
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Uracil, ≥99.0%
Supelco
Isobarbituric acid, analytical standard
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Barbituric acid, ReagentPlus®, 99%