Merck
  • Design, synthesis and biological evaluation of novel pyrenyl derivatives as anticancer agents.
Design, synthesis and biological evaluation of novel pyrenyl derivatives as anticancer agents.
European journal of medicinal chemistry (2014-12-03)
Debasish Bandyopadhyay, Jorge L Sanchez, Adrian M Guerrero, Fang-Mei Chang, Jose C Granados, John D Short, Bimal K Banik
ABSTRACT
Polycyclic aromatic hydrocarbons are widespread in nature with a toxicity range from non-toxic to extremely toxic. A series of pyrenyl derivatives has been synthesized following a four-step strategy where the pyrene nucleus is attached with a basic heterocyclic moiety through a carbon linker. Virtual screening of the physicochemical properties and druggability has been carried out. The cytotoxicity of the compounds (1-8) have been evaluated in vitro against a small panel of human cancer cell lines which includes two liver cancer (HepG2 and Hepa 1-6), two colon cancer (HT-29 and Caco-2) and one each for cervical (HeLa) and breast (MCF-7) cancer cell lines. The IC50 data indicate that compound 6 and 8 are the most effective cytotoxic agents in the present set of pyrenyl derivatives, suggesting that having a 4-carbon linker is more effective than a 5-carbon linker and the presence of amide carbonyl groups in the linker severely reduces the efficacy of the compound. The compounds showed selectivity toward cancer cells at lower doses (<5 μM) when compared with the normal hepatocytes. The mechanism of action supports the cell death through apoptosis in a caspase-independent manner without cleavage of poly (ADP-ribose) polymerase (PARP), even though the compounds cause plasma membrane morphological changes. The compounds, whether highly cytotoxic or mildly cytotoxic, localize to the membrane of cells. The compounds with either a piperidine ring (6) or an N-methyl piperazine (8) in the side chain were both capable of circumventing the drug resistance in SKOV3-MDR1-M6/6 ovarian cancer cells overexpressing P-glycoprotein. Qualitative structure-activity relationship has also been studied.
Sigma-Aldrich
Succinic anhydride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Isobutyl chloroformate, 98%
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Glutaric anhydride, 95%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water