Skip to Content
Merck
All Photos(1)

Key Documents

01934

Supelco

Dimethyl sulfoxide

for inorganic trace analysis, ≥99.99995% (metals basis)

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.24
grade:
for inorganic trace analysis
Assay:
≥99.9% (GC)
≥99.99995% (metals basis)
application(s):
sample preparation
bp:
189 °C (lit.)
189 °C
vapor pressure:
0.42 mmHg ( 20 °C)
0.42 mmHg

grade

for inorganic trace analysis

Quality Level

vapor density

2.7 (vs air)
2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)
0.42 mmHg

Assay

≥99.9% (GC)
≥99.99995% (metals basis)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F
42 %

color

colorless

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)
189 °C

mp

16-19 °C (lit.)
16-19 °C

solubility

H2O: miscible (completely)

density

1.099-1.101 g/mL at 20 °C
1.10 g/mL (lit.)

cation traces

Ag: ≤0.5 μg/kg
Al: ≤1 μg/kg
As: ≤0.01 mg/kg
Au: ≤0.5 μg/kg
Be: ≤0.5 μg/kg
Bi: ≤0.5 μg/kg
Ca: ≤20 μg/kg
Cd: ≤0.5 μg/kg
Ce: ≤0.5 μg/kg
Co: ≤0.5 μg/kg
Cr: ≤5 μg/kg
Cs: ≤0.5 μg/kg
Cu: ≤5 μg/kg
Fe: ≤20 μg/kg
Ga: ≤0.5 μg/kg
Ge: ≤0.5 μg/kg
Hg: ≤5 μg/kg
In: ≤10 μg/kg
Ir: ≤0.5 μg/kg
K: ≤10 μg/kg
Li: ≤0.5 μg/kg
Mg: ≤10 μg/kg
Mn: ≤5 μg/kg
Mo: ≤0.5 μg/kg
Na: ≤50 μg/kg
Ni: ≤5 μg/kg
Os: ≤1 μg/kg
Pb: ≤0.5 μg/kg
Pd: ≤0.5 μg/kg
Pt: ≤0.5 μg/kg
Rb: ≤0.5 μg/kg
Rh: ≤0.5 μg/kg
Ru: ≤0.5 μg/kg
Sb: ≤0.5 μg/kg
Se: ≤0.01 mg/kg
Sn: ≤0.5 μg/kg
Sr: ≤0.5 μg/kg
Ti: ≤5 μg/kg
Tl: ≤0.5 μg/kg
U: ≤0.5 μg/kg
V: ≤0.5 μg/kg
Zn: ≤5 μg/kg
Zr: ≤0.5 μg/kg

SMILES string

CS(C)=O

application(s)

sample preparation

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Enzymatic/chemical analysis of dietary fiber.: Focused on the analytical techniques for analyzing dietary fibers, which could be adapted for the analysis of complex organic matrices in the presence of DMSO ( Theander et al., 1994).

  • Levans: I. Fractionation, solution viscosity, and chemical analysis of levan produced by Streptococcus salivarius.: This research details the fractionation and analysis of bacterial levan, which could inform studies involving the solubilization and analysis of similar polysaccharides in DMSO ( Ehrlich et al., 1975).

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Other Notes

For information on dimethyl sulfoxide miscibility, please visit the following link:
Dimethyl Sulfoxide Miscibility/Immiscibility Table

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phillip McDonagh et al.
Veterinary microbiology, 174(3-4), 438-447 (2014-12-04)
Feline infectious peritonitis (FIP), a feline coronavirus (FCoV) induced disease, is almost invariably fatal with median life expectancy measured in days. Current treatment options are, at best, palliative. The objectives of this study were to evaluate a panel of nineteen
Claudia C Faria et al.
Cancer research, 75(1), 134-146 (2014-11-14)
Medulloblastoma is the most common malignant pediatric brain tumor, with metastases present at diagnosis conferring a poor prognosis. Mechanisms of dissemination are poorly understood and metastatic lesions are genetically divergent from the matched primary tumor. Effective and less toxic therapies
S L Khaw et al.
Leukemia, 28(6), 1207-1215 (2014-01-10)
Overexpression of the prosurvival protein Bcl-2 marks many B-lymphoid malignancies and contributes to resistance to many commonly used chemotherapeutic agents. The first effective BH3 mimetic inhibitors of Bcl-2, ABT-737 and navitoclax, also target Bcl-xL, causing dose-limiting thrombocytopenia. This prompted the
A Hillarp et al.
Journal of thrombosis and haemostasis : JTH, 12(9), 1545-1553 (2014-06-27)
Apixaban is an oral direct factor Xa inhibitor developed for the prophylaxis and treatment of thromboembolic disorders. Laboratory monitoring is not necessary, but the effects on common coagulation reagents and assays constitute clinically valuable information. To investigate the effects of apixaban
Rory W Blackler et al.
British journal of pharmacology, 172(4), 992-1004 (2014-10-10)
Hydrogen sulphide is an important mediator of gastrointestinal mucosal defence. The use of non-steroidal anti-inflammatory drugs (NSAIDs) is significantly limited by their toxicity in the gastrointestinal tract. Particularly concerning is the lack of effective preventative or curative treatments for NSAID-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service