09685
Ammonium ion solution for ISE
1000 mg/kg N, analytical standard (for ion-selective electrodes)
Synonym(s):
Ammonium chloride solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
Recommended Products
grade
analytical standard (for ion-selective electrodes)
for ion-selective electrodes
Quality Level
shelf life
limited shelf life, expiry date on the label
concentration
1000 mg/kg N
density
1.00 g/mL at 20 °C
0.995 g/mL at 25 °C (lit.)
SMILES string
[Cl-].[H][N+]([H])([H])[H]
InChI
1S/ClH.H3N/h1H;1H3
InChI key
NLXLAEXVIDQMFP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Visit our Sensor Applications portal to learn more.
Preparation Note
prepared with NH4Cl and H2O
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Cyrill A Rentsch et al.
European urology, 66(4), 677-688 (2014-03-29)
Whether the commonly used bacillus Calmette-Guérin (BCG) strains Connaught and Tice confer different treatment responses in non-muscle-invasive bladder cancer (NMIBC) is unknown. To compare clinical efficacy, immunogenicity, and genetics of BCG Connaught and Tice. A prospective randomized single-institution trial with
Christine Burkard et al.
PloS one, 9(7), e101762-e101762 (2014-07-16)
Studies of viral entry into host cells often rely on the detection of post-entry parameters, such as viral replication or the expression of a reporter gene, rather than on measuring entry per se. The lack of assays to easily detect
Allison L Boyd et al.
The Journal of experimental medicine, 211(10), 1925-1935 (2014-09-03)
Allogeneic hematopoietic stem cell (HSC) transplantation (HSCT) is currently the leading strategy to manage acute myeloid leukemia (AML). However, treatment-related morbidity limits the patient generalizability of HSCT use, and the survival of leukemic stem cells (LSCs) within protective areas of
Pontus Lundberg et al.
The Journal of experimental medicine, 211(11), 2213-2230 (2014-10-08)
The majority of patients with myeloproliferative neoplasms (MPNs) carry a somatic JAK2-V617F mutation. Because additional mutations can precede JAK2-V617F, it is questioned whether JAK2-V617F alone can initiate MPN. Several mouse models have demonstrated that JAK2-V617F can cause MPN; however, in
Kornelis S M van der Geest et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(7), 1927-1938 (2014-03-14)
Several lines of evidence indicate that B cells may be involved in the immunopathology of giant cell arteritis (GCA) and polymyalgia rheumatica (PMR). This study was undertaken to examine the distribution of defined B cell subsets, including effector B (Beff)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service