Merck
Olfactory proxy detection of dietary antioxidants in Drosophila.
Current biology : CB (2015-01-27)
Hany K M Dweck, Shimaa A M Ebrahim, Abu Farhan, Bill S Hansson, Marcus C Stensmyr
ABSTRACT
Dietary antioxidants play an important role in preventing oxidative stress. Whether animals in search of food or brood sites are able to judge the antioxidant content, and if so actively seek out resources with enriched antioxidant content, remains unclear. We show here that the vinegar fly Drosophila melanogaster detects the presence of hydroxycinnamic acids (HCAs)-potent dietary antioxidants abundant in fruit-via olfactory cues. Flies are unable to smell HCAs directly but are equipped with dedicated olfactory sensory neurons detecting yeast-produced ethylphenols that are exclusively derived from HCAs. These neurons are housed on the maxillary palps, express the odorant receptor Or71a, and are necessary and sufficient for proxy detection of HCAs. Activation of these neurons in adult flies induces positive chemotaxis, oviposition, and increased feeding. We further demonstrate that fly larvae also seek out yeast enriched with HCAs and that larvae use the same ethylphenol cues as the adults but rely for detection upon a larval unique odorant receptor (Or94b), which is co-expressed with a receptor (Or94a) detecting a general yeast volatile. We also show that the ethylphenols act as reliable cues for the presence of dietary antioxidants, as these volatiles are produced--upon supplementation of HCAs--by a wide range of yeasts known to be consumed by flies. For flies, dietary antioxidants are presumably important to counteract acute oxidative stress induced by consumption or by infection by entomopathogenic microorganisms. The ethylphenol pathway described here adds another layer to the fly's defensive arsenal against toxic microbes.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Methanol, SAJ special grade
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%