Merck
A cell model for the initial phase of sporadic Alzheimer's disease.
Journal of Alzheimer's disease : JAD (2014-06-06)
Carola Stockburger, Vicki A M Gold, Thea Pallas, Natalie Kolesova, Davide Miano, Kristina Leuner, Walter E Müller
ABSTRACT
Recent data suggest that the combined effect of oxidative stress due to aging and slightly elevated amyloid-β (Aβ) levels initiate Alzheimer's disease (AD) long before the clinical onset. Investigations of this early phase are hampered by the lack of cellular or animal models reflecting this scenario. We used SH-SY5Y cells stably transfected with an additional copy of the human AβPP gene and artificial aging by complex I inhibition. These cells show slightly elevated Aβ levels, moderately decreased ATP levels, impaired mitochondrial membrane potential, and decreased mitochondrial respiration. Assessing mitochondrial dynamics with three different methods reveals a distinct shift toward mitochondrial fission and fragmentation in SH-SY5Y AβPPwt cells. We also performed electron cryo-tomography of isolated mitochondria to reveal that there were no major differences between SH-SY5Y control and SH-SY5Y AβPPwt mitochondria with respect to swelling or loss of cristae. Dystrophic neurites are an early pathological feature of AD. Interestingly, SH-SY5Y AβPPwt cells exhibit significantly longer neurites, likely due to substantially elevated levels of sAβPPα. Complex I inhibition also shows substantial effects on mitochondrial dynamics, impairs neuritogenesis, and elevates Aβ levels in both cell types. In SH-SY5Y AβPPwt cells, these defects were more pronounced due to a relatively elevated Aβ and a reduced sAβPPα production. Our findings suggest that the progression from low Aβ levels to the beginning of AD takes place in the presence of oxidative stress during normal aging. This mechanism not only results from additive effects of both mechanisms on mitochondrial function but might also be additionally aggravated by altered amyloidogenic processing.
Sigma-Aldrich
Digitonin, ~50% (TLC)
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
SAFC
BIS-TRIS
Sigma-Aldrich
Anti-FIS1 antibody produced in rabbit
SAFC
BIS-TRIS
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Anti-DNM1L antibody produced in rabbit, affinity isolated antibody