Skip to Content
Merck
All Photos(9)

Documents

SAB2702049

Sigma-Aldrich

Anti-FIS1 antibody produced in rabbit

Synonym(s):

TTC11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

17 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 500-10000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FIS1(51024)

Immunogen

Recombinant fragment corresponding to a region within amino acids 1 and 152 of TTC11 (Uniprot ID#Q9Y3D6)

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:2000, IHC-P: 1:100-1:1000, WB: 1:500-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

The balance between fission and fusion regulates the morphology of mitochondria. TTC11 is a component of a mitochondrial complex that promotes mitochondrial fission (James et al., 2003 [PubMed 12783892]).[supplied by OMIM]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Yu et al.
PeerJ, 4, e2659-e2659 (2016-11-12)
Anesthetic postconditioning is a cellular protective approach whereby exposure to a volatile anesthetic renders a tissue more resistant to subsequent ischemic/reperfusion event. Sevoflurane postconditioning (SPostC) has been shown to exert cardioprotection against ischemia/reperfusion injury, but the underlying mechanism is unclear.
Estelle Balan et al.
Frontiers in physiology, 10, 1088-1088 (2019-09-12)
This study investigated whether regular endurance exercise maintains basal mitophagy and mitochondrial function during aging. Mitochondrial proteins and total mRNA were isolated from vastus lateralis biopsies (n = 33) of young sedentary (YS), old sedentary (OS), young active (YA), and
Bo Li et al.
Journal of neurochemistry, 154(5), 502-518 (2020-03-08)
Small extracellular vesicles (EVs), including exosomes, play multiple physiological roles. In neurodegenerative diseases, EVs can be pivotal in dispersing neuropathogenic proteins. This study investigates the role of neural stem cell (NSC)-derived EVs in a transgenic (Tg) mouse model of Alzheimer's
Carola Stockburger et al.
Journal of Alzheimer's disease : JAD, 42(2), 395-411 (2014-06-06)
Recent data suggest that the combined effect of oxidative stress due to aging and slightly elevated amyloid-β (Aβ) levels initiate Alzheimer's disease (AD) long before the clinical onset. Investigations of this early phase are hampered by the lack of cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service