Merck
  • The critical role of didodecyldimethylammonium bromide on physico-chemical, technological and biological properties of NLC.
The critical role of didodecyldimethylammonium bromide on physico-chemical, technological and biological properties of NLC.
Colloids and surfaces. B, Biointerfaces (2014-06-16)
C Carbone, A Campisi, D Manno, A Serra, M Spatuzza, T Musumeci, R Bonfanti, G Puglisi
ABSTRACT
Exploiting the experimental factorial design and the potentiality of Turbiscan AG Station, we developed and characterized unmodified and DDAB-coated NLC prepared by a low energy organic solvent free phase inversion temperature technique. A 22 full factorial experimental design was developed in order to study the effects of two independent variables (DDAB and ferulic acid) and their interaction on mean particle size and zeta potential values. The factorial planning was validated by ANOVA analysis; the correspondence between the predicted values of size and zeta and those measured experimentally confirmed the validity of the design and the equation applied for its resolution. The DDAB-coated NLC were significantly affected in their physico-chemical properties by the presence of DDAB, as showed by the results of the experimental design. The coated NLC showed higher physical stability with no particles aggregation compared to the unmodified NLC, as demonstrated by Turbiscan(®) AGS measurements. X-ray diffraction, Raman spectroscopy and Cryo-TEM images allowed us to assert that DDAB plays a critical role in increasing the lipids structural order with a consequent enhancement of the NLC physical stability. Furthermore, the results of the in vitro biological studies allow the revisiting of the role of DDAB to the benefit of glioblastoma treatment, due to its efficacy in increasing the NLC uptake and reducing the viability of human glioblastoma cancer cells (U87MG).
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Acetic acid solution, 1 M, 1 N
Sigma-Aldrich
Acetic acid, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.7%
Sigma-Aldrich
Acetic acid, JIS special grade, ≥99.7%
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Cetyl palmitate 15, European Pharmacopoeia (EP) Reference Standard
Supelco
Cetyl Palmitate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Didecyldimethylammonium bromide, 98%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Didodecyldimethylammonium bromide, 98%
Sigma-Aldrich
Methanol, JIS special grade, ≥99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, SAJ first grade, ≥99.5%
Sigma-Aldrich
Methanol, JIS 300, ≥99.8%, for residue analysis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, suitable for HPLC
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%