Merck
  • Loss of N-Myc interactor promotes epithelial-mesenchymal transition by activation of TGF-β/SMAD signaling.
Loss of N-Myc interactor promotes epithelial-mesenchymal transition by activation of TGF-β/SMAD signaling.
Oncogene (2013-06-19)
D J Devine, J W Rostas, B J Metge, S Das, M S Mulekar, J A Tucker, W E Grizzle, D J Buchsbaum, L A Shevde, R S Samant
ABSTRACT
Epithelial-mesenchymal transition is one of the critical cellular programs that facilitate the progression of breast cancer to an invasive disease. We have observed that the expression of N-myc interactor (NMI) decreases significantly during progression of breast cancer, specifically in invasive and metastatic stages. Recapitulation of this loss in breast cell lines with epithelial morphology (MCF10A (non-tumorigenic) and T47D (tumorigenic)) by silencing NMI expression causes mesenchymal-like morphological changes in 3D growth, accompanied by upregulation of SLUG and ZEB2 and increased invasive properties. Conversely, we found that restoring NMI expression attenuated the mesenchymal attributes of metastatic breast cancer cells, accompanied by distinctly circumscribed 3D growth with basement membrane deposition and decreased invasion. Further investigations into the downstream signaling modulated by NMI revealed that NMI expression negatively regulates SMAD signaling, which is a key regulator of cellular plasticity. We demonstrate that NMI blocks TGF-β/SMAD signaling via upregulation of SMAD7, a negative feedback regulator of the pathway. We also provide evidence that NMI activates STAT signaling, which negatively modulates TGF-β/SMAD signaling. Taken together, our findings suggest that loss of NMI during breast cancer progression could be one of the driving factors that enhance the invasive ability of breast cancer by aberrant activation of TGF-β/SMAD signaling.
Sigma-Aldrich
Anti-ZEB2 antibody produced in rabbit, affinity isolated antibody
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Monoclonal Anti-NMI antibody produced in mouse, clone 9D8, purified immunoglobulin, buffered aqueous solution
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anti-Laminin-5 Antibody, clone D4B5, Alexa Fluor 488 conjugated, clone D4B5, Chemicon®, from mouse
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material