Skip to Content
Merck
All Photos(2)

Key Documents

AV33694

Sigma-Aldrich

Anti-ZEB2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Zeb2 Antibody, Zeb2 Antibody - Anti-ZEB2 antibody produced in rabbit, Anti-KIAA0569, Anti-SIP-1, Anti-SIP1, Anti-SMADIP1, Anti-ZFHX1B, Anti-Zinc finger E-box binding homeobox 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

136 kDa

species reactivity

guinea pig, rat, human, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZEB2(9839)

General description

Zinc finger E-box-binding homeobox 2 (ZEB2) also known as Sip1 is a 137 kDa protein located in the nucleus where it functions as a DNA-binding transcriptional repressor that interacts with activated SMADs. Mutations in ZEB2 result in development of Mowat Wilson syndrome.

Immunogen

Synthetic peptide directed towards the N terminal region of human ZFHX1B

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

The ZFHX1B gene is a member of the delta-EF1/Zfh1 family of 2-handed zinc finger/homeodomain proteins. ZFHX1B is strongly transcribed at an early stage in the developing peripheral and central nervous systems of both mice and humans, in all neuronal regions of the brains of 25-week human fetuses and adult mice, and in numerous nonneural tissues.The SMADIP1 gene (also known as SIP1) is a member of the delta-EF1 (ZEB1; MIM 189909)/Zfh1 family of 2-handed zinc finger/homeodomain proteins. SMADIP1 interacts with receptor-mediated, activated full-length SMADs (see MIM 605568) (Verschueren et al., 1999 [PubMed 10400677]).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-58 AB056507.1 1-58 59-553 AL118674.1 57-551 554-5558 AB056507.1 553-5557 5559-5583 AI858477.1 1-25 c

Sequence

Synthetic peptide located within the following region: NVVDTGSETDEEDKLHIAEDDGIANPLDQETSPASVPNHESSPHVSQALL

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Mowat et al.
Journal of medical genetics, 40(5), 305-310 (2003-05-15)
MWS is a multiple congenital anomaly syndrome, first clinically delineated by Mowat et al in 1998. Over 45 cases have now been reported. All patients have typical dysmorphic features in association with severe intellectual disability, and nearly all have microcephaly
K Verschueren et al.
The Journal of biological chemistry, 274(29), 20489-20498 (1999-07-10)
Activation of transforming growth factor beta receptors causes the phosphorylation and nuclear translocation of Smad proteins, which then participate in the regulation of expression of target genes. We describe a novel Smad-interacting protein, SIP1, which was identified using the yeast
Damian Medici et al.
Nature medicine, 16(12), 1400-1406 (2010-11-26)
Mesenchymal stem cells can give rise to several cell types, but varying results depending on isolation methods and tissue source have led to controversies about their usefulness in clinical medicine. Here we show that vascular endothelial cells can transform into
D J Devine et al.
Oncogene, 33(20), 2620-2628 (2013-06-19)
Epithelial-mesenchymal transition is one of the critical cellular programs that facilitate the progression of breast cancer to an invasive disease. We have observed that the expression of N-myc interactor (NMI) decreases significantly during progression of breast cancer, specifically in invasive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service