Merck
  • Farnesyltransferase inhibitor, tipifarnib, prevents galactosamine/lipopolysaccharide-induced acute liver failure.
Farnesyltransferase inhibitor, tipifarnib, prevents galactosamine/lipopolysaccharide-induced acute liver failure.
Shock (Augusta, Ga.) (2014-07-22)
Kazuhiro Shirozu, Shuichi Hirai, Tomokazu Tanaka, Shinsuke Hisaka, Masao Kaneki, Fumito Ichinose
ABSTRACT
Acute liver failure (ALF) is a fatal syndrome associated with massive hepatocyte death. There is no cure for ALF except liver transplantation. Protein farnesylation is a lipid modification of cysteine residues that is catalyzed by farnesyltransferase (FTase) and has been proposed as an integral component of acute inflammation. Previously, we have demonstrated that FTase inhibitors improve survival in mouse models of endotoxemia and sepsis. Here we studied the effects of FTase inhibitor, tipifarnib, on galactosamine (GalN)/lipopolysaccharide (LPS)-induced ALF. The effects of tipifarnib (10 mg/kg, i.p.) were studied in GalN (400 mg/kg, i.p.)- and LPS (3 μg/kg)-challenged mice by histological and biochemical analyses. Galactosamine/LPS administration caused prominent liver injury characterized by the increased plasma alanine aminotransferase and aspartic aminotransferase levels, leading to significant mortality in mice. Tipifarnib inhibited GalN/LPS-induced caspase 3 activation, inflammatory cytokine production, and c-Jun N-terminal kinase phosphorylation in the liver. On the other hand, tipifarnib upregulated antiapoptotic protein, Bcl-xL, in the liver after GalN/LPS challenge. Tipifarnib also protected primary hepatocytes from GalN/tumor necrosis factor α-induced cell death by inhibiting caspase 3 activation and upregulating antiapoptotic proteins. Galactosamine/LPS-induced liver injury was associated with increased protein farnesylation in the liver. Tipifarnib prevented protein farnesylation in the liver and markedly attenuated liver injury and mortality in GalN/LPS-challenged mice. These results suggest that protein farnesylation is a novel potential molecular target to prevent hepatocyte death and acute inflammatory liver failure in fulminant hepatitis.
Sigma-Aldrich
Methyl Green, zinc chloride salt, certified by the Biological Stain Commission, Dye content 85 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, <0.5% crystal violet, Dye content 80 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, for microscopy (Bact., Bot., Hist.)
Sigma-Aldrich
Methyl Green, zinc chloride salt, ~85%
Sigma-Aldrich
Anti-Farnesyl Antibody, Chemicon®, from rabbit