Merck
Colloidal nanocrystals of lithiated group 14 elements.
Angewandte Chemie (International ed. in English) (2014-11-05)
Jacqueline E Cloud, Yonglong Wang, Tara S Yoder, Lauren W Taylor, Yongan Yang
ABSTRACT
The synthesis of colloidal nanocrystals (NCs) of lithiated group 14 elements (Z=Si, Ge, and Sn) is reported, which are Li4.4 Si, Li3.75 Si, Li4.4 Ge, and Li4.4 Sn. Lix Z compounds are highly reactive and cannot be synthesized by existing methods. The success relied on separating the surface protection from the crystal formation and using a unique passivating ligand. Bare Lix Z crystals were first produced by milling elemental Li and Z in an argon-filled jar. Then, under the assistance of additional milling, hexyllithium was added to passivate the freshly generated Lix Z NCs. This ball-milling-assisted surface protection method may be generalized to similar systems, such as Nax Z and Kx Z. Moreover, Li4.4 Si and Li4.4 Ge NCs were conformally encapsulated in carbon fibers, providing great opportunities for studying the potential of using Lix Z to mitigate the volume-fluctuation-induced poor cyclability problem confronted by Z anodes in lithium-ion batteries.
Sigma-Aldrich
N,N-Dimethylformamide, JIS special grade, ≥99.5%
Sigma-Aldrich
N,N-Dimethylformamide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Hexyllithium solution, 2.3 M in hexane
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Supelco
N,N-Dimethylformamide, analytical standard
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
Triethylsilane, 99%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Triethylsilane, 97%
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Hexane, suitable for HPLC
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Argon-40Ar, 99.95 atom %
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)