All Photos(1)
About This Item
Linear Formula:
40Ar
CAS Number:
Molecular Weight:
39.96
MDL number:
UNSPSC Code:
12142004
PubChem Substance ID:
Recommended Products
isotopic purity
99.95 atom %
Assay
≥99.5% (Total Argon)
<0.5% (Total others)
form
gas
impurities
<0.5% Total others
mass shift
depleted
SMILES string
[40Ar]
InChI
1S/Ar/i1+0
InChI key
XKRFYHLGVUSROY-IGMARMGPSA-N
Related Categories
Packaging
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Compr. Gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Linlin Li et al.
Small (Weinheim an der Bergstrasse, Germany), 11(20), 2429-2436 (2015-01-22)
Herein, the controlled synthesis of 3D hierarchical films on carbon cloth (CC) in a high yield through a hydrothermal process and their high photocatalytic properties are reported. As representative examples, the obtained ZnIn2 S4 /CdIn2 S4 composites are composed of
Andrew Riddell et al.
Methods (San Diego, Calif.), 82, 64-73 (2015-03-10)
Sorting performance can be evaluated with regard to Purity, Yield and/or Recovery of the sorted fraction. Purity is a check on the quality of the sample and the sort decisions made by the instrument. Recovery and Yield definitions vary with
Kali Janes et al.
Brain, behavior, and immunity, 44, 91-99 (2014-09-16)
Many commonly used chemotherapeutics including oxaliplatin are associated with the development of a painful chemotherapy-induced peripheral neuropathy (CIPN). This dose-limiting complication can appear long after the completion of therapy causing a significant reduction in quality-of-life and impeding cancer treatment. We
Yoshimitsu Sagara et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(33), 10397-10403 (2014-07-22)
Molecular assemblies that change photoluminescence color in response to thermal or mechanical stimulation without dissociation into the monomeric states in water are described herein. A dumbbell-shaped amphiphilic compound forms micellar molecular assemblies in water and exhibits yellow photoluminescence derived from
Godfrey J Quin et al.
Clinical & experimental ophthalmology, 43(2), 180-187 (2014-07-01)
Diabetic macular oedema (DMO) is the commonest cause of vision loss in people with diabetes. Laser photocoagulation can be effective in the treatment of DMO; however, its mechanism of action is still poorly understood. A better understanding of these mechanisms
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service