Merck
  • A novel pyrido-thieno-pyrimidine derivative activates p53 through induction of phosphorylation and acetylation in colorectal cancer cells.
A novel pyrido-thieno-pyrimidine derivative activates p53 through induction of phosphorylation and acetylation in colorectal cancer cells.
International journal of oncology (2014-10-24)
Mi Ae Kang, Mi-Sook Kim, Ji Young Kim, Young-Joo Shin, Jie-Young Song, Jae-Hoon Jeong
ABSTRACT
The tumor suppressor p53 plays a key role in regulation of the cell cycle, apoptosis and senescence in response to various stresses. We screened a library of 7920 chemical compounds for the p53 activator and identified N-[2-(dimethylamino)ethyl]-2,3-dimethyl-4-oxo-4H-pyrido[1,2-a]thieno[2,3-d]pyrimidine-9-carboxamide (PTP), which significantly increased p53-mediated reporter activity in colorectal cancer cells. PTP was found to induce p53 protein and activated transcription of downstream genes, such as p21 and PUMA, in HCT116 cells, leading to growth delay, G1-phase cell cycle arrest, cell senescence and cell death. Proximity ligation assay revealed that PTP weakened the interaction between p53 and murine double minute 2 (MDM2) in situ, thereby inhibiting MDM2-mediated p53 degradation. Although DNA damage has been known to promote phosphorylation of p53 and MDM2, thereby preventing their interaction and stabilizing p53, PTP did not cause DNA damage or activate any DNA damage response signaling. Instead, phosphorylation of p53 was mediated by Erk1/2 MAP kinase. In addition, PTP induced acetylation of p53 at Lys382 in a p300-dependent manner, but sirtuin (SIRT)1 and histone deacetylase (HDAC)1, a well-known p53-regulating deacetylase, were not involved. In the present study, the novel anticancer agent PTP was shown to cause the accumulation of p53 by inducing multiple post-translational modifications, as well as cell cycle arrest, senescence and cell death.
Sigma-Aldrich
Formaldehyde solution, JIS special grade, 36.0-38.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde solution, 10%
Sigma-Aldrich
Formaldehyde solution, SAJ first grade, ≥35.0%, contains methanol as stabilizer
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Glutaraldehyde solution, SAJ first grade, 20.0-26.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Anacardic acid
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
p-Terphenyl, suitable for scintillation, ≥98.5% (HPLC)
Supelco
p-Terphenyl, analytical standard
Sigma-Aldrich
p-Terphenyl, ≥99.5% (HPLC)