Merck
  • Sulfated glycosaminoglycan-based block copolymer: preparation of biocompatible chondroitin sulfate-b-poly(lactic acid) micelles.
Sulfated glycosaminoglycan-based block copolymer: preparation of biocompatible chondroitin sulfate-b-poly(lactic acid) micelles.
Biomacromolecules (2014-05-27)
André R Fajardo, Alexandre Guerry, Elizandra A Britta, Celso V Nakamura, Edvani C Muniz, Redouane Borsali, Sami Halila
ABSTRACT
Despite a growing interest in amphiphilic polysaccharide-based diblock copolymers as functional polymeric drug delivery nanosystems, biologically relevant sulfated glycosaminoglycan systems were not yet investigated. Here, we report the synthesis and the self-assembly properties in water of chondroitin sulfate-b-poly(lactic acid) (CS-b-PLA(n)). The CS-b-PLA(n) were synthesized using click-grafting onto method implying reducing-end alkynation of low-molecular weight depolymerized CS (M(w) = 5000 g·mol(-1)) and azide-terminated functionalization of PLAn (M(w) = 6500 g·mol(-1) (n = 46) and M(w) = 1700 g·mol(-1) (n = 20)). The diblock copolymer self-assembled in water giving rise to spherical micelles that were characterized in solution using dynamic/static light scattering and at dry state by TEM technique. In vitro assays on healthy cells showed that at high concentrations, up to 10 μg·mL(-1), CS-b-PLA(n) were noncytotoxic. Those preliminary studies are promising in the perspective to use them as biocompatible nanovehicles for anticancer drug delivery.
Sigma-Aldrich
Sodium azide, SAJ first grade, ≥97.0%
Sigma-Aldrich
Triethylamine, SAJ first grade, ≥98.0%
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
Triethylamine, ≥99.5%
Supelco
4-Nitrophenol, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Sigma-Aldrich
p-Toluenesulfonyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
p-Toluenesulfonyl chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%