Merck
  • Effects of LB broth, naphthalene concentration, and acetone on the naphthalene degradation activities by Pseudomonas putida G7.
Effects of LB broth, naphthalene concentration, and acetone on the naphthalene degradation activities by Pseudomonas putida G7.
Water environment research : a research publication of the Water Environment Federation (2015-01-30)
Su-Yun Chang, Xue-Gong Liu, Bi-Qiong Ren, Bo Liu, Kai Zhang, Honggui Zhang, Yao Wan
ABSTRACT
Luria-Bertani broth and acetone were usually used in naphthalene degradation experiments as nutrient and solvent. However, their effect on the degradation was seldom mentioned. In this work, we investigated the effect of LB, naphthalene concentration, and acetone on the degradation of naphthalene by Pseudomonas putida G7, which is useful for the degradation of naphthalene on future field remediation. By adding LB, the naphthalene degradation efficiencies and naphthalene dioxygenase were both decreased by 98%, while the catechol dioxygenase was decreased by 90%. Degradation of naphthalene was also inhibited when naphthalene concentration was 56 ppm and higher, which was accompanied with the accumulation of orange-colored metabolism products. However, acetone can stimulate the degradation of naphthalene, and the stimulation was more obvious when naphthalene concentration was lower than 2000 ppm. By assaying the enzyme activities of naphthalene dioxygenase and catechol dioxygenase, it was thought that the degradation efficiency was depending on the more sensitive enzymes on the complicated conditions.
Product Description
Supelco
Melting point standard 79-81°C, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, 99%
Supelco
Naphthalene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, for residue analysis, JIS 5000
Sigma-Aldrich
Acetone, for residue analysis, ≥99.5%
Sigma-Aldrich
Acetone, ≥99.5%, for residue analysis
Sigma-Aldrich
Acetone, for chromatography, ≥99.8%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetone, JIS special grade, ≥99.5%
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%