Skip to Content
Merck
All Photos(2)

Key Documents

91489

Supelco

Naphthalene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8
CAS Number:
Molecular Weight:
128.17
Beilstein:
1421310
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

vapor density

4.4 (vs air)

vapor pressure

0.03 mmHg ( 25 °C)

product line

TraceCERT®

autoignition temp.

978 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

5.9 %

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

218 °C (lit.)

mp

80-82 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

c1ccc2ccccc2c1

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H

InChI key

UFWIBTONFRDIAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Naphthalene is an aromatic hydrocarbon[1], found to be nonpolar in nature.[2] It is generally produced from petroleum or coal-tar.[1] Naphthalene, an organic compound, is widely used as an intermediate in chemical and plastic industry. Its other applications include, its role in the manufacturing of fungicides/insecticides[3] and as a model compound for investigating the metabolism of polycyclic aromatic hydrocarbons (PAHs) by bacteria. The metabolism of this compound by Polaromonas naphthalenivorans CJ2 via the gentisate pathway followed by respirometry, GC-MS and cell-free enzyme assay has been studied.[4]
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support. Naphthalene may be used as a certified reference material for the quantification of the analyte in honey samples using head-space solid-phase microextraction (HS-SPME) combined with gas chromatography–mass spectrometry (GC/MS) technique.[5]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

173.3 °F - closed cup

Flash Point(C)

78.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naphthalene metabolism and growth inhibition by naphthalene in Polaromonas naphthalenivorans strain CJ2
Pumphrey MG and Madsen LE
Microbiology, 153(11), 3730-3738 (2007)
Development of a rapid and sensitive method for the simultaneous determination of 1, 2-dibromoethane, 1, 4-dichlorobenzene and naphthalene residues in honey using HS-SPME coupled with GC?MS
Tsimeli K, et al.
Analytica Chimica Acta, 617, 64-71 (2008)
Introduction to Organic Laboratory Techniques: A Small Scale Approach (2005)
The 100 Most Important Chemical Compounds: A Reference Guide (2007)
Determination of naphthalene metabolites in human urine by liquid chromatography?mass spectrometry with electrospray ionization
Andreoli R, et al.
Journal of Chromatography A, 847(1-2), 9-17 (1999)

Articles

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service