Skip to Content
Merck
All Photos(1)

Key Documents

40053

Supelco

Naphthalene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
7822574
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

c1ccc2ccccc2c1

InChI

1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H

InChI key

UFWIBTONFRDIAS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Sigma-Aldrich offers an extensive line of Fluka and Supelco brand neats and single-component solutions for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyses. Naphthalene solution may be prepared, via dissolving polycyclic aromatic hydrocarbons in methanol.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative SERS sensors for environmental analysis of naphthalene
Peron.O, et al.
Analyst, 135, 1018-1022 (2011)
K Vijayavel et al.
Biomedical and environmental sciences : BES, 26(4), 295-302 (2013-03-29)
To investigate protective effect of Coleus aromaticus leaf extract against naphthalene induced hepatotoxicity in rats. Eighteen male rats were divided into three groups. Group I rats were treated as control. Group II rats were intraperitoneally administered with naphthalene (435 mg/kg
Qianqian Shi et al.
Journal of environmental sciences (China), 25(1), 188-194 (2013-04-17)
A high-surface-area carbon (KC-1) was prepared from waste polystyrene-based ion exchange resin by KOH activation and used for naphthalene adsorption. The carbon exhibited a good hydrophobic nature with developed porous structure, favoring the adsorption of organic compounds. The Brunauer-Emmett-Teller surface
Kaori Yamaguchi et al.
Journal of chromatography. A, 1288, 149-154 (2013-03-26)
Hydro gel formed by 5'-guanosine monophosphate (GMP) in the presence of a potassium ion is expected to exhibit interesting selectivity in capillary electrophoretic separations. Here, we estimated the conditional association constants between the hydro gel (G-gel) and aromatic compounds by
Xu-Liang Cao et al.
Journal of food protection, 75(12), 2163-2171 (2012-12-06)
A method based on isotope dilution headspace solid-phase microextraction, followed by gas chromatography and mass spectrometry, was developed for the determination of naphthalene in foods. Optimum method sensitivity was achieved by the addition of NaCl in water at saturation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service