Merck
  • Safety assessment of butylated hydroxyanisole and butylated hydroxytoluene as antioxidant food additives.
Safety assessment of butylated hydroxyanisole and butylated hydroxytoluene as antioxidant food additives.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1999-10-29)
G M Williams, M J Iatropoulos, J Whysner
ABSTRACT
Butylated hydroxyanisole (BHA) and butylated hydroxytoluene (BHT) are widely used antioxidant food additives. They have been extensively studied for potential toxicities. This review details experimental studies of genotoxicity and carcinogenicity which bear on cancer hazard assessment of exposure to humans. We conclude that BHA and BHT pose no cancer hazard and, to the contrary, may be anticarcinogenic at current levels of food additive use.
Product Description
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Butylhydroxyanisole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Sigma-Aldrich
Butylated hydroxyanisole, 99%, FCC, FG
Sigma-Aldrich
Butylated hydroxyanisole, ≥98.5%
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, SAJ first grade, ≥99.0%
Supelco
Butylated hydroxyanisole, analytical standard
Supelco
3,5-Di-tert-butyl-4-hydroxytoluene, analytical standard
Butylhydroxytoluene, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Supelco
Butylated hydroxyanisole, Pharmaceutical Secondary Standard; Certified Reference Material