Skip to Content
Merck
All Photos(1)

Key Documents

B1212000

Butylhydroxyanisole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Butylated hydroxyanisole, 2(3)-t-Butyl-4-hydroxyanisole, 2(3)-t-Butylhydroquinone monomethyl ether, BHA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3(OCH3)OH
CAS Number:
Molecular Weight:
180.24
EC Number:
MDL number:
UNSPSC Code:
41116107
E Number:
E320
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

6.2 (vs air)

API family

butylhydroxyanisole

autoignition temp.

599 °F

manufacturer/tradename

EDQM

mp

58-60 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

O(C)c1cc(c(cc1)O)C(C)(C)C

InChI

1S/C11H16O2/c1-11(2,3)9-7-8(13-4)5-6-10(9)12/h5-7,12H,1-4H3

InChI key

MRBKEAMVRSLQPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Butylhydroxyanisole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

241.9 °F - Pensky-Martens closed cup

Flash Point(C)

116.6 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuchaya Ngamsuk et al.
Foods (Basel, Switzerland), 8(9) (2019-09-07)
The effects of dry processing and maturity on antioxidant activity, total phenolic content, total procyanidins, and identity of phenolic compounds in coffee leaves were evaluated. Fresh coffee leaves were tray-dried at 40 °C for 8 h before total phenolic content
Dubravka Vitali Čepo et al.
Molecules (Basel, Switzerland), 23(8) (2018-08-22)
Waste remaining after the production of olive oil (olive pomace) is known to contain significant amounts of phenolic compounds that exert different types of biological activities, primarily acting as antioxidants. In this work, a sustainable approach that combines ultrasound-assisted extraction
Carolina Reyes-Contreras et al.
Environmental technology, 41(11), 1358-1365 (2018-10-12)
Organic micropollutants (OMP) in the household and industrial wastewater are not efficiently removed by conventional treatment processes and a significant fraction ends in sludge. Proper valorization technologies become fundamental to attain sustainable sewage sludge management, with anaerobic digestion (AD) as
Xiaoyan Hou et al.
Antioxidants (Basel, Switzerland), 9(1) (2020-01-16)
In order to evaluate the antioxidant and α-glucosidase activities of polysaccharides from Chrysanthemum morifolium cv. Hangju (CMPs), the response surface methodology was applied to optimize the parameters for extraction progress of CMPs by ultrasound, with heat reflex extraction (HRE) performed
Yann Malato et al.
Gastroenterology, 143(6), 1597-1608 (2012-08-28)
The transcription factor nuclear factor κB (NF-κB) is activated by the IκB kinase complex. The regulatory subunit of this complex, NF-κB essential modifier (NEMO or IKBKG), is a tumor suppressor. Hepatocyte-specific deletion of NEMO induces chronic liver inflammation that leads

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service