Skip to Content
Merck
All Photos(1)

Key Documents

03-4720

Sigma-Aldrich

2,6-Di-tert-butyl-4-methylphenol

SAJ first grade, ≥99.0%

Synonym(s):

2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Beilstein:
1911640
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

grade

SAJ first grade

vapor density

7.6 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥99.0%

autoignition temp.

878 °F

availability

available only in Japan

bp

265 °C (lit.)

mp

69-73 °C (lit.)

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gen-ichiro Arimura et al.
Plant physiology, 146(3), 965-973 (2008-01-01)
Continuous mechanical damage initiates the rhythmic emission of volatiles in lima bean (Phaseolus lunatus) leaves; the emission resembles that induced by herbivore damage. The effect of diurnal versus nocturnal damage on the initiation of plant defense responses was investigated using
Butylated hydroxytoluene (BHT): a review.
H Babich
Environmental research, 29(1), 1-29 (1982-10-01)
Metabolism and pulmonary toxicity of butylated hydroxytoluene (BHT).
H Witschi et al.
Pharmacology & therapeutics, 42(1), 89-113 (1989-01-01)
G M Williams et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 37(9-10), 1027-1038 (1999-10-29)
Butylated hydroxyanisole (BHA) and butylated hydroxytoluene (BHT) are widely used antioxidant food additives. They have been extensively studied for potential toxicities. This review details experimental studies of genotoxicity and carcinogenicity which bear on cancer hazard assessment of exposure to humans.
A M Malkinson
Environmental mutagenesis, 5(3), 353-362 (1983-01-01)
Although the average American's daily consumption of BHT can be measured in milligrams, there are numerous reports that BHT causes organ damage in laboratory animals. Only a few genotoxic effects of BHT have been reported, however, including mutagenicity in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service