Merck
  • A membrane bioreactor for the simultaneous treatment of acetone, toluene, limonene and hexane at trace level concentrations.
A membrane bioreactor for the simultaneous treatment of acetone, toluene, limonene and hexane at trace level concentrations.
Water research (2013-03-19)
R Lebrero, D Volckaert, R Pérez, R Muñoz, H Van Langenhove
ABSTRACT
The performance of a flat-membrane biofilm reactor (MBR) for the removal of acetone, toluene, limonene and hexane at concentrations ranging from 1.3 to 3.2 mg m(-3) was investigated at different gas residence times (GRT): 60, 30, 15 and 7 s. A preliminary abiotic test was conducted to assess the mass transport of the selected volatile organic compounds (VOCs) through the membrane. A reduced transport of limonene and hexane was observed with water present over the dense side of the membrane. The presence of a biofilm attached on the dense side of the membrane following bioreactor inoculation significantly increased VOC transport. High acetone and toluene removals (>93%) were recorded in the MBR regardless of the GRT. To remediate the low hexane removal performance (RE < 24%) recorded at the initial stages of the process, a re-inoculation of the membrane with a hexane-degrading consortium embedded in silicon oil was performed. Although hexane removal did not exceed 27%, this re-inoculation increased limonene removals up to 90% at a GRT of 7 s. The absence of inhibition of hexane biodegradation by substrate competition confirmed that hexane removal in the MBR was indeed limited by the mass transfer through the membrane. Despite the low carbon source spectrum and load, the microbiological analysis of the communities present in the MBR showed high species richness (Shannon-Wiener indices of 3.2-3.5) and a high pair-wise similarity (84-97%) between the suspended and the attached biomass.
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Hexane, JIS special grade, ≥96.0%
Sigma-Aldrich
Hexane, for residue analysis, JIS 5000
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Hexane, JIS 1000, ≥96.0%, for residue analysis
Sigma-Aldrich
Toluene, JIS 300, for residue analysis, ≥99.8%
Sigma-Aldrich
Hexane, SAJ first grade, ≥95.0%
Sigma-Aldrich
Toluene, JIS 1000, for residue analysis, ≥99.8%
Sigma-Aldrich
Hexane, JIS 300, ≥96.0%, for residue analysis
Sigma-Aldrich
Toluene, JIS special grade, ≥99.5%
Sigma-Aldrich
Hexane, ≥96.0%, suitable for residual phthalate analysis
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hexane-1-13C, 99 atom % 13C
Sigma-Aldrich
Hexane, suitable for HPLC
Supelco
Toluene, analytical standard
Supelco
Hexane, analytical standard
Sigma-Aldrich
(S)-(−)-Limonene, 96%
Supelco
(S)-(−)-Limonene, analytical standard
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
(S)-(−)-Limonene, ≥95%, FG
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%