Merck
  • Flexible minerals: self-assembled calcite spicules with extreme bending strength.
Flexible minerals: self-assembled calcite spicules with extreme bending strength.
Science (New York, N.Y.) (2013-03-16)
Filipe Natalio, Tomas P Corrales, Martin Panthöfer, Dieter Schollmeyer, Ingo Lieberwirth, Werner E G Müller, Michael Kappl, Hans-Jürgen Butt, Wolfgang Tremel
ABSTRACT
Silicatein-α is responsible for the biomineralization of silicates in sponges. We used silicatein-α to guide the self-assembly of calcite "spicules" similar to the spicules of the calcareous sponge Sycon sp. The self-assembled spicules, 10 to 300 micrometers (μm) in length and 5 to 10 μm in diameter, are composed of aligned calcite nanocrystals. The spicules are initially amorphous but transform into calcite within months, exhibiting unusual growth along [100]. They scatter x-rays like twinned calcite crystals. Whereas natural spicules evidence brittle failure, the synthetic spicules show an elastic response, which greatly enhances bending strength. This remarkable feature is linked to a high protein content. With nano-thermogravimetric analysis, we measured the organic content of a single spicule to be 10 to 16%. In addition, the spicules exhibit waveguiding properties even when they are bent.
Sigma-Aldrich
Calcium carbonate, CP
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Marble, CP