Skip to Content
Merck
All Photos(3)

Documents

900500

Sigma-Aldrich

Calcium carbonate

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

product line

ReagentPlus®
Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

pH

8

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium carbonate is a calcium salt commonly found in shells and skeleton of marine animals. It is an inexpensive, non-toxic and easily available reagent with a wide range of applications such as uptake of CO2 and extraction of heavy metals and arsenic from ground water. Its crystals exist as 3 different polymorphs namely, calcite, aragonite, and vaterite. The kinetics of pyrolysis of calcium carbonate under non-isothermal conditions have been investigated.

Application

Calcium carbonate may be used:
  • As a pore generating agent to prepare porous ceramics.
  • As chelometric standard for the standardization of EDTA solution.
  • To extract aluminium by microwave induced thermal activation of coal fly ash (CFA).
Iron-Catalyzed Efficient Synthesis of Amides from Aldehydes and Amine Hydrochloride Salts

Iron-Catalyzed Efficient Synthesis of Amides from Aldehydes and Amine Hydrochloride Salts

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of calcium carbonate decomposition.
Rao TR.
Chemical Engineering & Technology, 19(4), 373-377 (1996)
Polymorphs calcium carbonate on temperature reaction.
Chong KY, et al.
AIP Conference Proceedings, 1614(1) (2014)
Permeability of porous ceramic based on calcium carbonate as pore generating agent.
Sim?o L, et al.
Ceramics International, 41(3), 4782-4788 (2015)
Kanwaljit Kaur
Handbook of Water and Wastewater Analysis, 247-248 (2007)
Opposite particle size effect on amorphous calcium carbonate crystallization in water and during heating in air.
Zou Z, et al.
Chemistry of Materials, 27(12), 4237-4246 (2015)

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service