21067
Calcium carbonate
reference material for titrimetry, certified by BAM, ≥99.5%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
reference material
Quality Level
Assay
≥99.5%
quality
certified by BAM
technique(s)
titration: suitable
solubility
H2O: insoluble
density
2.93 g/mL at 25 °C (lit.)
format
neat
SMILES string
[Ca++].[O-]C([O-])=O
InChI
1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2
InChI key
VTYYLEPIZMXCLO-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Calcium carbonate volumetric standard is manufactured under stringent conditions and measured with the highest possible precision. The standard is qualified by volumetric complexometric titration with photometric endpoint detection. Its certified value and uncertainty measurement are in accordance with ISO Guide 35.
Application
It finds its use as a reference material for the standardization of volumetric solutions in complexometric titrations.
Features and Benefits
- Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
- High-quality offering accurate titer determinations
- Accompanied by a certificate of analysis (CoA)
Analysis Note
Exact concentration, expiry date, traceability and detailed information on certification can be found on the certificate and certification report, included in each package. Content and expiry date on label.
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Science (New York, N.Y.), 339(6125), 1298-1302 (2013-03-16)
Silicatein-α is responsible for the biomineralization of silicates in sponges. We used silicatein-α to guide the self-assembly of calcite "spicules" similar to the spicules of the calcareous sponge Sycon sp. The self-assembled spicules, 10 to 300 micrometers (μm) in length
PloS one, 8(3), e58781-e58781 (2013-03-26)
An organic matrix consisting of a protein-polysaccharide complex is generally accepted as an important medium for the calcification process. While the role this "calcified organic matrix" plays in the calcification process has long been appreciated, the complex mixture of proteins
Journal of women's health (2002), 22(11), 915-929 (2013-10-18)
Clinical outcomes of the Women's Health Initiative (WHI) calcium plus vitamin D supplementation trial have been reported during 7.0 years of active intervention. We now report outcomes 4.9 years after the intervention stopped and cumulative findings. Postmenopausal women (N=36,282) were
Carbohydrate polymers, 97(2), 315-320 (2013-08-06)
Thermoplastic starch (TPS) was prepared using compression molding and chicken eggshell was used as a filler. The effect of the eggshell powder (EP) on the properties of TPS was compared with the effect of commercial calcium carbonate (CC). The organic
Lancet (London, England), 382(9900), 1268-1277 (2013-07-23)
Phosphate binders (calcium-based and calcium-free) are recommended to lower serum phosphate and prevent hyperphosphataemia in patients with chronic kidney disease, but their effects on mortality and cardiovascular outcomes are unknown. We aimed to update our meta-analysis on the effect of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service