Merck
  • Effects of formic acid and phytase supplementation on digestibility and use of phosphorus and zinc in growing pigs.
Effects of formic acid and phytase supplementation on digestibility and use of phosphorus and zinc in growing pigs.
Journal of animal science (2013-02-13)
R Blank, M Naatjes, C Baum, K Köhling, P Ader, U Roser, A Susenbeth
ABSTRACT
Two studies, arranged according to a 4 × 4 Latin square design, were conducted to assess effects of dietary acidification on fungal 3-phytase (PHY) efficacy in growing pigs. In Exp. 1, effects of supplementing 500 units/kg feed of PHY and 4.7 g/kg HCOOH either alone or in combination on the use of P and Zn in growing pigs fed a pelleted diet based on wheat (Triticum aestivum), barley (Hordeum vulgare), and soybean (Glycine max) meal were investigated. In Exp. 2 the same dietary treatments were fed except that PHY supplementation was increased to 1000 units/kg. In both experiments, PHY supplementation increased (P < 0.05) P digestibility and retention. A PHY × HCOOH supplementation interaction on P balance was observed (P < 0.05), indicating that the combination of the additives may increase P digestibility and retention. Effects of HCOOH and PHY on Zn use followed a similar pattern. Supplementation of 1000 units/kg of PHY further increased P and Zn retention compared to supplementation of 500 units/kg. In conclusion, the present study indicated that HCOOH supplementation to diets with microbial PHY may increase PHY efficacy.
Sigma-Aldrich
Formic acid, JIS special grade, ≥98.0%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Potassium formate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium formate-13C, 99 atom % 13C
Sigma-Aldrich
Potassium formate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium formate, 99.998% trace metals basis
Sigma-Aldrich
Sodium formate, BioUltra, ≥99.0% (NT)
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Sodium formate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Sodium formate, reagent grade, 97%
Sigma-Aldrich
Ammonium formate, SAJ first grade, ≥95.0%
Supelco
Calcium formate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Thallium(I) formate, 97%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Calcium formate, BioUltra, ≥99.0% (T)
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Cesium formate, 98%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium formate, reagent grade, 97%