Merck
  • Selective sorption of dissolved organic carbon compounds by temperate soils.
Selective sorption of dissolved organic carbon compounds by temperate soils.
PloS one (2012-12-05)
Sindhu Jagadamma, Melanie A Mayes, Jana R Phillips
ABSTRACT
Physico-chemical sorption onto soil minerals is one of the major processes of dissolved organic carbon (OC) stabilization in deeper soils. The interaction of DOC on soil solids is related to the reactivity of soil minerals, the chemistry of sorbate functional groups, and the stability of sorbate to microbial degradation. This study was conducted to examine the sorption of diverse OC compounds (D-glucose, L-alanine, oxalic acid, salicylic acid, and sinapyl alcohol) on temperate climate soil orders (Mollisols, Ultisols and Alfisols). Equilibrium batch experiments were conducted using 0-100 mg C L(-1) at a solid-solution ratio of 1∶60 for 48 hrs on natural soils and on soils sterilized by γ-irradiation. The maximum sorption capacity, Q(max) and binding coefficient, k were calculated by fitting to the Langmuir model. Ultisols appeared to sorb more glucose, alanine, and salicylic acid than did Alfisols or Mollisols and the isotherms followed a non-linear pattern (higher k). Sterile experiments revealed that glucose and alanine were both readily degraded and/or incorporated into microbial biomass because the observed Q(max) under sterile conditions decreased by 22-46% for glucose and 17-77% for alanine as compared to non-sterile conditions. Mollisols, in contrast, more readily reacted with oxalic acid (Q(max) of 886 mg kg(-1)) and sinapyl alcohol (Q(max) of 2031 mg kg(-1)), and no degradation was observed. The reactivity of Alfisols to DOC was intermediate to that of Ultisols and Mollisols, and degradation followed similar patterns as for Ultisols. This study demonstrated that three common temperate soil orders experienced differential sorption and degradation of simple OC compounds, indicating that sorbate chemistry plays a significant role in the sorptive stabilization of DOC.
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Oxalic acid, SAJ first grade, ≥97.0%
Supelco
Oxalic acid concentrate, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Oxalic acid solution, 0.05 M
Sigma-Aldrich
Oxalic acid solution, 0.5 M
Sigma-Aldrich
Sodium oxalate solution, 5 mM
Sigma-Aldrich
Sodium oxalate solution, 0.0125 M
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Sodium oxalate, BioXtra
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%