Skip to Content
Merck
All Photos(2)

Key Documents

139718

Sigma-Aldrich

Trimethylphenylammonium tribromide

97%

Synonym(s):

Phenyltrimethylammonium bromide dibromide, Phenyltrimethylammonium bromide-perbromide, Phenyltrimethylammonium tribromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(Br3)C6H5
CAS Number:
Molecular Weight:
375.93
Beilstein:
3919459
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

110-115 °C (dec.) (lit.)

SMILES string

Br[Br-]Br.C[N+](C)(C)c1ccccc1

InChI

1S/C9H14N.Br3/c1-10(2,3)9-7-5-4-6-8-9;1-3-2/h4-8H,1-3H3;/q+1;-1

InChI key

PRXNKYBFWAWBNZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Brominating reagent for 1,2-addition of bromine to the double bond of α,β-unsaturated compounds.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heteroatom Chem., 3, 345-345 (1992)
Tetrahedron, 50, 6433-6433 (1994)
A Ranz et al.
Journal of biochemical and biophysical methods, 69(1-2), 3-14 (2006-03-28)
In the present study, a derivatization method for the determination of acidic herbicides has been investigated. This procedure involves the methylation with the quaternary ammonium salt trimethylanilinium hydroxide (TMAH) directly in the gas chromatographic auto-sampler vial for analysis by gas
Alec N Salt
Advances in oto-rhino-laryngology, 59, 140-148 (2002-03-12)
The inner ear fluids are remarkably 'unstirred' so that it cannot be assumed that applied drugs are dispersed throughout the fluid spaces. Calculation of the effective concentrations achieved when drugs are applied directly to the inner ear is made possible
Cláudia Margalho et al.
Journal of analytical toxicology, 29(5), 309-313 (2005-08-18)
A simple and rapid method was validated to determine furosemide in whole blood. The experimental work was performed so that all validation parameters are considered simultaneously in a one-day assay protocol. A solid-phase extraction procedure using BondElut-LRC Certify columns was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service