Merck
Separation of lipid classes by HPLC on a cyanopropyl column.
Lipids (2011-11-22)
Petter Olsson, Jan Holmbäck, Bengt Herslöf
ABSTRACT
A new method for the separation and identification of lipid classes by normal-phase HPLC on a cyanopropyl column is described. The use of a simple binary gradient, with toluene as a component, provided a rapid separation of non-polar as well as phospholipid classes. The inherent small differences in performances between possible non-polar eluent components of the gradient, such as hexane, heptane, and iso-octane, had a pronounced impact on retention times for individual phospholipid classes. Separation of molecular species within a lipid class could also be observed.
Product Description
Supelco
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
2,2,4-Trimethylpentane, analytical standard
Sigma-Aldrich
2,2,4-Trimethylpentane, ACS reagent, ≥99.0%
Sigma-Aldrich
2,2,4-Trimethylpentane, suitable for HPLC, ≥99%
Sigma-Aldrich
2,2,4-Trimethylpentane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
Sigma-Aldrich
2,2,4-Trimethylpentane, JIS special grade
Sigma-Aldrich
2,2,4-Trimethylpentane, ≥99.5%, suitable for absorption spectrum analysis
Sigma-Aldrich
2,2,4-Trimethylpentane, SAJ first grade, ≥98.0%
Sigma-Aldrich
2,2,4-Trimethylpentane, ReagentPlus®, ≥99%
Sigma-Aldrich
2,2,4-Trimethylpentane, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%