Merck
Multiple DNA-binding sites in Tetrahymena telomerase.
Nucleic acids research (2008-01-05)
Sharon N Finger, Tracy M Bryan
ABSTRACT
Telomerase is a ribonucleoprotein enzyme that maintains chromosome ends through de novo addition of telomeric DNA. The ability of telomerase to interact with its DNA substrate at sites outside its catalytic centre ('anchor sites') is important for its unique ability to undergo repeat addition processivity. We have developed a direct and quantitative equilibrium primer-binding assay to measure DNA-binding affinities of regions of the catalytic protein subunit of recombinant Tetrahymena telomerase (TERT). There are specific telomeric DNA-binding sites in at least four regions of TERT (the TEN, RBD, RT and C-terminal domains). Together, these sites contribute to specific and high-affinity DNA binding, with a K(d) of approximately 8 nM. Both the K(m) and K(d) increased in a stepwise manner as the primer length was reduced; thus recombinant Tetrahymena telomerase, like the endogenous enzyme, contains multiple anchor sites. The N-terminal TEN domain, which has previously been implicated in DNA binding, shows only low affinity binding. However, there appears to be cooperativity between the TEN and RNA-binding domains. Our data suggest that different DNA-binding sites are used by the enzyme during different stages of the addition cycle.
Product Description
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® DNA LoBind tubes, capacity 2.0 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® Deepwell plates, DNA LoBind, 384 wells, white plate, conical bottom, colorless wells, capacity 200 μL, pack of 120 ea (10 bags × 12 plates)
Eppendorf® Protein LoBind Microplate 384/V-PP, PCR clean, V-bottom clear wells, white border, pack of 240 ea (10 bags × 24 plates)
Eppendorf® Deepwell plates, DNA LoBind, 384 wells, white plate, conical bottom, colorless wells, capacity 200 μL, pack of 40 ea (5 bags × 8 plates)
Eppendorf® DNA LoBind tubes, capacity 0.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® DNA LoBind tubes, capacity 1.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 120 ea (10 bags × 12 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Deepwell plates, DNA LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 1000 μL, pkg of 80 ea (10 bags × 8 plates)
Eppendorf® Protein LoBind tubes, capacity 1.5 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Protein LoBind tubes, capacity 2.0 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, yellow plate, conical bottom, colorless wells, capacity 500 μL, pkg of 40 ea (5 bags × 8 plates)
Eppendorf® Protein LoBind tubes, capacity 0.5 mL, PCR clean, pkg of 100 ea (2 x 50ea)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 80 ea (10 bags × 8 plates)
Eppendorf® Deepwell plates, Protein LoBind, 96 wells, yellow plate, conical bottom, colorless wells, capacity 500 μL, pkg of 20 ea (5 bags × 4 plates)