Skip to Content
Merck
All Photos(1)

Key Documents

EP022431021

Eppendorf® DNA LoBind tubes

capacity 1.5 mL, PCR clean, pkg of 250 ea (5 x 50ea)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103037

material

cap (push fit)
conical bottom
polypropylene
polypropylene cap

sterility

non-sterile

feature

DNase free
PCR clean
RCF 30,000 × g
RNase free
graduations

packaging

pkg of 250 ea (5 x 50ea)

manufacturer/tradename

Eppendorf® 022431021

capacity

1.5 mL

diam.

10.7 mm

color

clear

suitability

suitable for PCR

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf LoBind® tubes, snap cap, 1.5 mL, PCR clean, colorless, 250 tubes (5 bags × 50 tubes)
  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy up-scaling
  • Precise lid sealing for minimal evaporation rates in tube format
  • Rated up to 30,000 x g (25,000 x g for 2.0 mL) centrifugation speed for molecular biology applications
Maximize nucleic acid recovery. Eppendorf LoBind Tubes signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules. The ideal solution for sample preparation and long-term storage of your precious nucleic acids

Features and Benefits

Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf LoBind is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelly Hodge et al.
Journal of proteomics, 88, 92-103 (2013-03-19)
Mass spectrometry, in the past five years, has increased in speed, accuracy and use. With the ability of the mass spectrometers to identify increasing numbers of proteins the identification of undesirable peptides (those not from the protein sample) has also
Cláudia P Grou et al.
The Journal of biological chemistry, 283(21), 14190-14197 (2008-03-25)
According to current models of peroxisomal biogenesis, newly synthesized peroxisomal matrix proteins are transported into the organelle by Pex5p. Pex5p recognizes these proteins in the cytosol, mediates their membrane translocation, and is exported back into the cytosol in an ATP-dependent
Arzu Umar et al.
Proteomics, 7(2), 323-329 (2006-12-14)
Proteomics assays hold great promise for unraveling molecular events that underlie human diseases. Effective analysis of clinical samples is essential, but this task is considerably complicated by tissue heterogeneity. Laser capture microdissection (LCM) can be used to selectively isolate target
Hongchang Qu et al.
Immunobiology, 218(4), 496-505 (2012-07-17)
Therapeutic modulation of the complement system has become increasingly important in line with the growing recognition of the role of complement in numerous diseases. Compstatin, a peptidic inhibitor that acts at the central level of the complement cascade, is currently
Graziano Colombo et al.
Free radical biology & medicine, 52(9), 1584-1596 (2012-03-06)
Cigarette smoke, a complex mixture of over 7000 chemicals, contains many components capable of eliciting oxidative stress, which may induce smoking-related disorders, including oral cavity diseases. In this study, we investigated the effects of whole (mainstream) cigarette smoke on human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service