Skip to Content
Merck
All Photos(1)

Key Documents

EP951033308

Eppendorf® Deepwell plates, Protein LoBind, 96 wells

white plate, conical bottom, colorless wells, capacity 500 μL, pkg of 80 ea (10 bags × 8 plates)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
material:
clear wells
colorless wells
conical bottom
polypropylene
white plate
size:
96 well
sterility:
non-sterile
binding type:
low binding surface
feature:
lid: no

material

clear wells
colorless wells
conical bottom
polypropylene
white plate

sterility

non-sterile

feature

lid: no

packaging

pkg of 80 ea (10 bags × 8 plates)

manufacturer/tradename

Eppendorf® 951033308

capacity

500 μL

size

96 well

well volume

1000 μL

color

white border

suitability

suitable for (protein analysis)
suitable for PCR

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Deepwell Plate 96/1000 µL, Protein LoBind], wells colorless, 1,000 µL, PCR clean, white, 80 plates (10 bags × 8 plates)
  • Eppendorf LoBind material ensures excellent sample recovery for improved assay results
  • Free of surface coating (e.g., silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • Unique OptiTrack® matrix: 30 % faster sample identification and fewer pipetting errors
  • RecoverMax well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing in plates

Features and Benefits

  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • High-contrast Unique OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors
  • RecoverMax® well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelly Hodge et al.
Journal of proteomics, 88, 92-103 (2013-03-19)
Mass spectrometry, in the past five years, has increased in speed, accuracy and use. With the ability of the mass spectrometers to identify increasing numbers of proteins the identification of undesirable peptides (those not from the protein sample) has also
Arzu Umar et al.
Proteomics, 7(2), 323-329 (2006-12-14)
Proteomics assays hold great promise for unraveling molecular events that underlie human diseases. Effective analysis of clinical samples is essential, but this task is considerably complicated by tissue heterogeneity. Laser capture microdissection (LCM) can be used to selectively isolate target
Byung-Gyu Kim et al.
Proteomics, 6(4), 1166-1174 (2006-01-20)
Runx2 is a key transcription factor in osteoblast differentiation, and its activity is regulated by fibroblast growth factors (FGFs). Craniosynostosis, characterized by premature suture closure, results from mutations that generate constitutively active FGF receptors (FGFRs). We previously showed that FGF/FGFR-activated
Steven J Bark et al.
Journal of proteome research, 6(11), 4511-4516 (2007-09-14)
Differential recovery of peptides due to nonspecific adsorption can seriously compromise reproducibility and quality of proteomic data for peptide analyses by liquid chromatography-mass spectrometry (LC-MS). This study demonstrates large variations in reproducibility and quantitation of LC-MS data for peptides derived
Hongchang Qu et al.
Immunobiology, 218(4), 496-505 (2012-07-17)
Therapeutic modulation of the complement system has become increasingly important in line with the growing recognition of the role of complement in numerous diseases. Compstatin, a peptidic inhibitor that acts at the central level of the complement cascade, is currently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service